Skip to Content
Merck
All Photos(1)

Key Documents

543365

Sigma-Aldrich

IR-820

Dye content 80 %

Synonym(s):

2-[2-[2-Chloro-3-[[1,3-dihydro-1,1-dimethyl-3-(4-sulfobutyl)-2H-benzo[e]indol-2-ylidene]-ethylidene]-1-cyclohexen-1-yl]-ethenyl]-1,1-dimethyl-3-(4-sulfobutyl)-1H-benzo[e]indolium hydroxide inner salt, sodium salt, New Indocyanine Green

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H50ClN2NaO6S2
CAS Number:
Molecular Weight:
849.47
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

composition

Dye content, 80%

Quality Level

mp

>300 °C (lit.)

λmax

820 nm

SMILES string

[Na+].CC1(C)\C(=C\C=C2/CCCC(\C=C\C3=[N+](CCCCS([O-])(=O)=O)c4ccc5ccccc5c4C3(C)C)=C2Cl)N(CCCCS([O-])(=O)=O)c6ccc7ccccc7c16

InChI

1S/C46H51ClN2O6S2.Na/c1-45(2)40(48(28-9-11-30-56(50,51)52)38-24-20-32-14-5-7-18-36(32)42(38)45)26-22-34-16-13-17-35(44(34)47)23-27-41-46(3,4)43-37-19-8-6-15-33(37)21-25-39(43)49(41)29-10-12-31-57(53,54)55;/h5-8,14-15,18-27H,9-13,16-17,28-31H2,1-4H3,(H-,50,51,52,53,54,55);/q;+1/p-1

InChI key

RANIQVAJHXBIAY-UHFFFAOYSA-M

General description

IR-820 is an indocyanine dye, which has a broad absorption band at 690 nm in aqueous solution. It is a near infrared dye (NIR) with a molar absorptivity of 72 x 104 dm3mol−1cm−1.

Application

IR-820 is a cost-effective imaging agent with low cytotoxicity. It can be used for image guided photothermal therapy for the treatment of cancer. It is also used as a contrasting agent for the detection and quantification of infected tissues in animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Near-infrared imaging of injured tissue in living subjects using IR-820
Prajapati SI, et al.
Molecular Imaging, 8(1), 7290-2009 (2009)
SERS characterization of the indocyanine-type dye IR-820 on gold and silver nanoparticles in the near infrared
Neves TBV and Andrade GFS
Journal of Spectroscopy (New York, NY, United States), 2015 (2015)
IR 820 stabilized multifunctional polycaprolactone glycol chitosan composite nanoparticles for cancer therapy
Kumar P and Srivastava R
Royal Society of Chemistry Advances, 5(69), 56162-56170 (2015)
IR 820 dye encapsulated in polycaprolactone glycol chitosan: Poloxamer blend nanoparticles for photo immunotherapy for breast cancer
Kumar P and Srivastava R
Materials Science and Engineering, C, 57(69), 321-327 (2015)
Xia Dong et al.
Biomaterials, 209, 111-125 (2019-04-30)
As synergistic photothermal immunotherapy has developed as one of the most attractive strategies for cancer therapy, it is crucial to design an effective photothermal immunotherapy system to enhance the synergistic anti-tumor effect and reveal the essential role of each treatment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service