Skip to Content
Merck
All Photos(2)

Key Documents

537691

Sigma-Aldrich

2,6-Dimethyl-4-heptanone

new

technical grade

Synonym(s):

Diisobutyl ketone, Isovalerone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCH2CH(CH3)2
CAS Number:
Molecular Weight:
142.24
Beilstein:
1743163
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

vapor density

4.9 (vs air)

vapor pressure

1.7 mmHg ( 20 °C)

autoignition temp.

745 °F

composition

  • 2,6-Dimethyl-4-heptanone (main component)
  • 4,6-Dimethyl-2-heptanone (secondary component)
  • 3,3,5-Trimethylcyclohexanone (minor component)

expl. lim.

0.8-6.2 %, 100 °F

refractive index

n20/D 1.412 (lit.)

bp

165-170 °C (lit.)

density

0.808 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CC(C)CC(=O)CC(C)C

InChI

1S/C9H18O/c1-7(2)5-9(10)6-8(3)4/h7-8H,5-6H2,1-4H3

InChI key

PTTPXKJBFFKCEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Dimethyl-4-heptanone (di-i-butyl ketone) is widely employed as an industrial solvent. It undergoes gas-phase reaction with the OH radical in the presence of NOx to afford acetone and 2-methylpropanal. 2,6-Dimethyl-4-heptanone is formed as a product during the thermal degradation of Yttrium(III) isovalerate Y.

Application

2,6-Dimethyl-4-heptanone may be employed as a diluent for the isolation of pentanedioic acid by using Amberlite LA-2.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Products of the gas-phase OH radical-initiated reactions of 4-methyl-2-pentanone and 2, 6-dimethyl-4-heptanone.
Atkinson R and Aschmann SM.
International Journal of Chemical Kinetics, 27(3), 261-275 (1993)
The Vapor Pressures and Some Other Properties of Di-i-butyl Ketone and Di-i-butylcarbinol.
Stross FH, et al.
Journal of the American Chemical Society, 69(7), 1629- 1630 (1947)
Thermal decomposition of Yttrium (III) isovalerate in argon.
Grivel JC, et al.
Journal of Analytical and Applied Pyrolysis (2016)
Extractive Separation of Pentanedioic Acid by Amberlite LA-2 in Various Solvents.
Uslu H, et al.
Journal of Chemical and Engineering Data (2016)
Marte Balchen et al.
Analytica chimica acta, 716, 16-23 (2012-01-31)
A totally new concept based on electrokinetic migration was evaluated for the extraction of three biologically active peptides from human plasma. Angiotensin 2, leu-enkephalin, and endomorphin 1 migrated from a diluted human plasma sample (2 mL, positive electrode), through a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service