Skip to Content
Merck
All Photos(1)

Key Documents

537306

Sigma-Aldrich

D-Hydroorotic acid

98%

Synonym(s):

R-2,6-Dioxohexahydro-4-pyrimidinecarboxylic acid, Dihydro-D-orotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O4
CAS Number:
Molecular Weight:
158.11
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

optical activity

[α]20/D −33°, c = 1 in NaHCO3

mp

255 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)[C@H]1CC(=O)NC(=O)N1

InChI

1S/C5H6N2O4/c8-3-1-2(4(9)10)6-5(11)7-3/h2H,1H2,(H,9,10)(H2,6,7,8,11)/t2-/m1/s1

InChI key

UFIVEPVSAGBUSI-UWTATZPHSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mihwa Lee et al.
Journal of molecular biology, 348(3), 523-533 (2005-04-14)
Escherichia coli dihydroorotase has been crystallized in the presence of the product, L-dihydroorotate (L-DHO), and the structure refined at 1.9A resolution. The structure confirms that previously reported (PDB entry 1J79), crystallized in the presence of the substrate N-carbamyl-D,L-aspartate (D, L-CA-asp)
Claudia A McDonald et al.
Biochemistry, 50(14), 2714-2716 (2011-03-16)
Adding the two residues comprising the conserved proton-transfer network of Class 2 dihydroorotate dehydrogenase (DHOD) to the Cys130Ser Class 1A DHOD did not restore the function of the active site base or rapid flavin reduction. Studies of triple, double, and
Michelle L Zaharik et al.
Microbiology (Reading, England), 153(Pt 8), 2472-2482 (2007-07-31)
Mutants capable of utilizing the pyrimidine biosynthetic intermediates carbamoylaspartate and dihydroorotate for growth were derived from pyrimidine auxotrophs of Salmonella enterica serovar Typhimurium LT2. The gain-of-function phenotypes both resulted from mutations in a single gene, yhiT, the third gene of
Rebecca L Fagan et al.
Biochemistry, 45(50), 14926-14932 (2006-12-13)
Dihydroorotate dehydrogenases (DHODs) oxidize dihydroorotate (DHO) to orotate using the FMN prosthetic group to abstract a hydride equivalent from C6 and a protein residue (Ser for Class 2 DHODs) to deprotonate C5. The fundamental question of whether the scission of
Mark A Anderson et al.
Biochemistry, 45(23), 7132-7139 (2006-06-07)
In the pyrimidine biosynthetic pathway, N-carbamyl-L-aspartate (CA-asp) is converted to L-dihydroorotate (DHO) by dihydroorotase (DHOase). The mechanism of this important reaction was probed using primary and secondary 15N and 13C isotope effects on the ring opening of DHO using isotope

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service