Skip to Content
Merck
All Photos(2)

Key Documents

531278

Sigma-Aldrich

Poly(isobutylene-alt-maleic anhydride)

average Mw ~6,000, 12-200 mesh (85%)

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H2O3 · C4H8)X
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

average Mw ~6,000

Quality Level

particle size

12-200 mesh (85%)

inherent viscosity

0.45±0.05 dL/g, in DMF(30 °C)(lit.)

transition temp

Tg (DSC) 141 °C (onset)

solubility

aqueous base: soluble

density

1.3 g/mL at 25 °C (lit.)

SMILES string

O1C(=O)C=CC1=O.CC(=C)C

InChI

1S/C4H2O3.C4H8/c5-3-1-2-4(6)7-3;1-4(2)3/h1-2H;1H2,2-3H3

InChI key

RPOCFUQMSVZQLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(isobutylene-alt-maleic anhydride) was used to prepare amphiphilic polymer with dodecylamine. The polymer is a dispersing and surface active agent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Relaxation times of colloidal iron platinum in polymer matrixes.
Morales MP, et al.
Journal of Materials Chemistry, 19(35), 6381-6386 (2009)
Xueyan Mu et al.
Carbohydrate polymers, 90(4), 1582-1586 (2012-09-05)
In the present study, we report a facile method to fabricate polyelectrolyte based core-shell nanofibers with the assistance of the high gradient electric potential between the tip of capillary and the collector. The core-shell structure and the composition of each
P B Kernoff et al.
Blood, 63(1), 31-41 (1984-01-01)
Circulating antibodies to factor VIII (anti-VIII, "inhibitors") occurring in patients with hemophilia neutralize porcine factor VIII less readily than human factor VIII in vitro. Over an 18-mo period, 8 patients with anti-VIII were treated with 45 courses (297 infusions) of
Abhimanyu Paraskar et al.
Nanotechnology, 22(26), 265101-265101 (2011-05-18)
The use of cisplatin, a first line chemotherapy for most cancers, is dose-limited due to nephrotoxicity. While this toxicity can be addressed through nanotechnology, previous attempts at engineering cisplatin nanoparticles have been limited by the impact on the potency of
R D Jayant et al.
Drug delivery, 16(6), 331-340 (2009-07-18)
Alginate microspheres loaded with dexamethasone were prepared by the droplet generator technique. Important parameters affecting drug release, including initial drug content, the type of polyelectrolyte coating, and a combination of different ratios of coated and uncoated microspheres were investigated to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service