Skip to Content
Merck
All Photos(2)

Key Documents

531081

Sigma-Aldrich

N-Hydroxynaphthalimide triflate

electronic grade, ≥99%

Synonym(s):

N-Hydroxynaphthalimide trifluoromethanesulfonate, HNT, NHN-TF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H6F3NO5S
CAS Number:
Molecular Weight:
345.25
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

grade

electronic grade

Assay

≥99%

mp

212.1-214.2 °C

solubility

PGMEA: 1%
ethyl lactate: <1%
γ-butyrolactone: ~2%

SMILES string

FC(F)(F)S(=O)(=O)ON1C(=O)c2cccc3cccc(C1=O)c23

InChI

1S/C13H6F3NO5S/c14-13(15,16)23(20,21)22-17-11(18)8-5-1-3-7-4-2-6-9(10(7)8)12(17)19/h1-6H

InChI key

LWHOMMCIJIJIGV-UHFFFAOYSA-N

General description

N-Hydroxynaphthalimide triflate (HNT) is a photoacid generator(PAG) that forms an acid upon exposure to UV light which can be used to generate patterned structures. It can also be used to tune the solubility of a polymer by forming acid linkages.

Application

HNT can be used to form patterned nanoparticles that find potential applications in volumetric manufacturing of semiconductor devices. It can also be used in the photo-responsive pore reopening of zeolites that facilitates the regulation of the gas permeation process. UV-irradiated trimethylsilyl ultrathin cellulose based films can be formed in presence of HNT as a PAG.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photoregeneration of trimethylsilyl cellulose as a tool for microstructuring ultrathin cellulose supports
Wolfberger A, et al.
Molecules (Basel), 19(10), 16266-16273 (2014)
Jan Kendall de Kruif et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 101, 90-102 (2016-02-07)
Newly discovered active macromolecules are highly promising for therapy, but poor bioavailability hinders their oral use. Microencapsulation approaches, such as protein prilling into microspheres, may enable protection from gastrointestinal (GI) enzymatic degradation. This would increase bioavailability mainly for local delivery
Rajeev N Bahuguna et al.
Physiologia plantarum, 159(1), 59-73 (2016-08-16)
High night temperature (HNT) is a major constraint to sustaining global rice production under future climate. Physiological and biochemical mechanisms were elucidated for HNT-induced grain yield and quality loss in rice. Contrasting rice cultivars (N22, tolerant; Gharib, susceptible; IR64, high
Surface modification of zeolites using benzene-1, 4-diboronic acid to form gated micropores with mild and photo responsive pore reopening
Fujiwara M, et al.
Chemical Engineering Journal, 146(3), 520-526 (2009)
Jayabrata Maity et al.
Carbohydrate polymers, 182, 159-171 (2017-12-28)
Composite type adsorbent was prepared by integrating chitosan (Cs) with crosslinked polymethacrylic acid (PMA) and nano sized halloysite nanotube (HNT). The structure of the resulting Cs-PMA/HNT adsorbents was characterized by FTIR, NMR, XRD, TGA, SEM/EDX and rheological properties. These functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service