Skip to Content
Merck
All Photos(2)

Key Documents

525693

Sigma-Aldrich

6-Bromo-2-pyridinecarboxaldehyde

97%

Synonym(s):

2-Bromo-6-formylpyridine, 2-Bromopyridine-6-carboxaldehyde, 2-Bromopyridyl-6-carboxaldehyde, 2-Formyl-6-bromopyridine, 6-Bromo-2-formylpyridine, 6-Bromo-2-pyridinecarbaldehyde, 6-Bromopicolinaldehyde, 6-Bromopyridine-2-aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BrNO
CAS Number:
Molecular Weight:
186.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

81-85 °C (lit.)

functional group

aldehyde
bromo

storage temp.

2-8°C

SMILES string

[H]C(=O)c1cccc(Br)n1

InChI

1S/C6H4BrNO/c7-6-3-1-2-5(4-9)8-6/h1-4H

InChI key

QWFHFNGMCPMOCD-UHFFFAOYSA-N

General description

6-Bromo-2-pyridinecarboxaldehyde is a pyridine derivative. It participates in the synthesis of meso-substituted trans-A2B2-porphyrin.

Application

6-Bromo-2-pyridinecarboxaldehyde may be used in the synthesis of:
  • 6-([2,2′-bi(1,3-dithiolylidene)]-4-yl)picolinaldehyde
  • tris(2-pyridylmethyl)amine (Tpy) derivative having Tpy core bearing a 3-formyl-phenyl substituent on one of the three arms
  • syn- and anti- forms of 4-(6-bromopyridin-2-yl)-3-methyl-3-phenyloxetan-2-one
  • 6-(2,4,6-triisopropylphenyl)-2-pyridinecarboxaldehyde
Substrate used in a study of a rhodium-catalyzed, reductive aldol coupling with divinyl ketones leading to syn ß-hydroxyenones.
Useful building block for Tris[(pyridyl)methyl]amine ligands.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magdalena M Makowska-Grzyska et al.
Inorganic chemistry, 42(23), 7472-7488 (2003-11-11)
Divalent manganese, cobalt, nickel, and zinc complexes of 6-Ph(2)TPA (N,N-bis((6-phenyl-2-pyridyl)methyl)-N-((2-pyridyl)methyl)amine; [(6-Ph(2)TPA)Mn(CH(3)OH)(3)](ClO(4))(2) (1), [(6-Ph(2)TPA)Co(CH(3)CN)](ClO(4))(2) (2), [(6-Ph(2)TPA)Ni(CH(3)CN)(CH(3)OH)](ClO(4))(2) (3), [(6-Ph(2)TPA)Zn(CH(3)CN)](ClO(4))(2) (4)) and 6-(Me(2)Ph)(2)TPA (N,N-bis((6-(3,5-dimethyl)phenyl-2-pyridyl)methyl)-N-((2-pyridyl)methyl)amine; [(6-(Me(2)Ph)(2)TPA)Ni(CH(3)CN)(2)](ClO(4))(2) (5) and [(6-(Me(2)Ph)(2)TPA)Zn(CH(3)CN)](ClO(4))(2) (6)) have been prepared and characterized. X-ray crystallographic characterization of 1A.CH(3)()OH and 1B.2CH(3)()OH (differing solvates
NHC-Promoted asymmetric β-lactone formation from arylalkylketenes and electron-deficient benzaldehydes or pyridinecarboxaldehydes.
Douglas J, et al.
The Journal of Organic Chemistry, 78(8), 3925-3938 (2013)
Awatef Ayadi et al.
Beilstein journal of organic chemistry, 11, 1379-1391 (2015-10-02)
The synthesis and full characterization of two tetrathiafulvalene-appended azine ligands, namely 2-([2,2'-bi(1,3-dithiolylidene)]-4-yl)-6-((2,4-dinitrophenyl)hydrazono)methyl)pyridine (L1) and 5-([2,2'-bi(1,3-dithiolylidene)]-4-yl)-2-((2,4-dinitrophenyl)hydrazono)methyl)pyridine (L2) are described. The crystal structure of ligand L1 indicates that the ligand is completely planar with the presence of a strong intramolecular N3-H3···O1 hydrogen
Francesca A Scaramuzzo et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(49), 16809-16813 (2013-11-01)
Reliable determination of the enantiomeric excess of free amino acids can be obtained by measuring the induced circular dichroism of a multicomponent assembly formed by a modified tris(2-pyridylmethyl)amine ligand, a zinc salt, and the amino acid of interest. The systems
Steric and Electronic Effects in the Formation and Carbon Disulfide Reactivity of Dinuclear Nickel Complexes Supported by Bis (iminopyridine) Ligands.
Bheemaraju A, et al.
Organometallics, 32(10), 2952-2962 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service