Skip to Content
Merck
All Photos(2)

Key Documents

525472

Sigma-Aldrich

D-Phenylalanine methyl ester hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NH2)CO2CH3 · HCL
CAS Number:
Molecular Weight:
215.68
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

reaction type: solution phase peptide synthesis

mp

159-163 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

Cl[H].COC(=O)[C@H](N)Cc1ccccc1

InChI

1S/C10H13NO2.ClH/c1-13-10(12)9(11)7-8-5-3-2-4-6-8;/h2-6,9H,7,11H2,1H3;1H/t9-;/m1./s1

InChI key

SWVMLNPDTIFDDY-SBSPUUFOSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J S Shin et al.
Biotechnology and bioengineering, 69(5), 577-583 (2000-07-18)
We recently demonstrated (J Am Chem Soc 121:3334-3340, 1999) that enzymatic enantioselectivity in organic solvents can be markedly enhanced by temporarily enlarging the substrate via salt formation. In the present study, this approach was expanded by finding that, in addition
Ikuo Kira et al.
Journal of bioscience and bioengineering, 108(3), 190-193 (2009-08-12)
Screening was carried out for microorganisms able to produce N-(l-alpha-l-aspartyl)-l-phenylalanine methyl ester [APM] from l-isoasparagine and l-phenylalanine methyl ester hydrochloride. Of the 422 strains examined, 44 strains belonging to the family Enterobacteriaceae were found to produce APM. The enzyme catalyzing
Shotaro Tsuchiyama et al.
Biotechnology progress, 23(4), 820-823 (2007-05-08)
The PST-01 protease is a metalloprotease that has zinc ion at the active center and is very stable in the presence of water-soluble organic solvents. The reaction rates and the equilibrium yields of the aspartame precursor N-carbobenzoxy-L-aspartyl-L-phenylalanine methyl ester (Cbz-Asp-Phe-OMe)
L Ye et al.
Journal of molecular recognition : JMR, 11(1-6), 75-78 (1999-03-17)
We have studied the possibility of shifting a thermodynamically unfavourable enzymatic equilibrium towards product formation via the addition of a highly specific adsorbent. The commercially interesting enzymatic condensation of Z-L-aspartic acid with L-phenylalanine methyl ester to the sweetener aspartame was
Y Watanabe et al.
Biochimica et biophysica acta, 1337(1), 40-46 (1997-01-04)
A newly isolated actinomycete belonging to Saccharothrix sp. was found to produce a unique enzyme catalyzing D-amino acid transfer. The enzyme, which was tentatively named D-amino acid transferase, was purified 2600-fold to electrophoretic homogeneity and the molecular mass was 41

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service