Skip to Content
Merck
All Photos(1)

Key Documents

523429

Sigma-Aldrich

2,3-Dibromo-1,4-naphthoquinone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H4Br2O2
CAS Number:
Molecular Weight:
315.95
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

mp

218-222 °C (lit.)

functional group

bromo
ketone

SMILES string

BrC1=C(Br)C(=O)c2ccccc2C1=O

InChI

1S/C10H4Br2O2/c11-7-8(12)10(14)6-4-2-1-3-5(6)9(7)13/h1-4H

InChI key

PSMABVOYZJWFBV-UHFFFAOYSA-N

General description

2,3-Dibromo-1,4-naphthoquinone is a 2,3-disubstituted 1,4-naphthoquinone.[1] It is a plumbagin derivative and an acaricide.[2] It undergoes photochemical reaction with 2-methoxy-1-alkene to yield derivatives of 2-(2-alkanonyl)-1,4-naphthoquinone.[1]

Application

2,3-Dibromo-1,4-naphthoquinone may be used in the synthesis of 3-[3-(2-carboxy-ethylsulfanyl)-1,4-dioxo-1,4-dihydro-naphthalen-2-ylsulfanyl]-propionic acid[3] and NSC 95397 (a protein tyrosine phosphatase antagonist).[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A one-pot synthesis of 1, 4-naphthoquinone-2, 3-bis-sulfides catalysed by a commercial laccase.
Wellington KW, et al.
Green Chemistry, 14(9), 2567-2576 (2012)
Vincent P Peyregne et al.
Molecular cancer therapeutics, 4(4), 595-602 (2005-04-14)
Cdc25 phosphatases are important in cell cycle control and activate cyclin-dependent kinases (Cdk). Efforts are currently under way to synthesize specific small-molecule Cdc25 inhibitors that might have anticancer properties. NSC 95397, a protein tyrosine phosphatase antagonist from the National Cancer
Chi-Hoon Lee et al.
Journal of microbiology and biotechnology, 18(2), 314-321 (2008-03-01)
Acaricidal effects of materials derived from Diospyros kaki roots against Dermatophagoides farinae and D. pteronyssinus were assessed using impregnated fabric disk bioassay and compared with that of the commercial benzyl benzoate. The observed responses varied according to dosage and mite
A new method of ?-keto alkyl chain introduction into 1, 4-naphthoquinone.
Maruyama K, et al.
Chemistry Letters (Jpn), 13(3), 371-374 (1984)
Xiao-Fei Shang et al.
Scientific reports, 8(1), 1609-1609 (2018-01-27)
As important secondary plant metabolites, naphthoquinones exhibit a wide range of biological activities. However, their potential as sustainable alternatives to synthetic acaricides has not been studied. This study for the first time investigates the acaricidal activity of naphthoquinones against Psoroptes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service