Skip to Content
Merck
All Photos(1)

Key Documents

519324

Sigma-Aldrich

1-(tert-Butyldimethylsilyloxy)-1-methoxyethene

97%

Synonym(s):

(1,1-Dimethylethyl)[(1-methoxyethenyl)oxy]dimethylsilane, (tert-Butyl)[(1-methoxyethenyl)oxy]dimethylsilane, 1-Methoxy-1-(tert-butyldimethylsiloxy)ethylene, 1-Methoxy-1-[(tert-butyldimethylsilyl)oxy]ethene, 1-Methoxy-1-[(tert-butyldimethylsilyl)oxy]ethylene, Ketene methyl t-butyldimethylsilyl acetal, Ketene tert-butyldimethylsilyl methyl acetal, tert-Butyl[(1-methoxyvinyl)oxy]dimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSi(CH3)2OC(OCH3)=CH2
CAS Number:
Molecular Weight:
188.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.429 (lit.)

bp

62 °C/9 mmHg (lit.)

density

0.863 g/mL at 25 °C (lit.)

SMILES string

COC(=C)O[Si](C)(C)C(C)(C)C

InChI

1S/C9H20O2Si/c1-8(10-5)11-12(6,7)9(2,3)4/h1H2,2-7H3

InChI key

UVCCWXJGWMGZAB-UHFFFAOYSA-N

Application

1-(tert-Butyldimethylsilyloxy)-1-methoxyethene may be used in the following
  • Synthesis of β-hydroxy esters via Mukaiyama Aldol reaction with heterocyclic aldehydes in the presence of Lewis acid.
  • Asymmetric Mukaiyama-Mannich reaction with isatin-derived ketimines to synthesize indolyl-β3, 3-aminoacid esters.
  • Broadrup′s synthesis of epothilone D.
  • Preparation of N-benzylidene benzylamine N-oxide.
  • Synthesis of nitroso acetals.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New indenyl phosphinooxazoline complexes of iron and their catalytic activity in the Mukaiyama aldol reaction.
Lenze M, et al.
Tetrahedron Letters, 51(21), 2855-2858 (2010)
Enantioselective addition of ketene silyl acetals to nitrones catalyzed by chiral titanium complexes. Synthesis of optically active ?-amino acids.
Murahashi SI, et al.
Journal of the American Chemical Society, 124(12), 2888-2889 (2002)
Recent developments in the syntheses of the epothilones and related analogues.
Watkins EB, et al.
European Journal of Organic Chemistry, 18, 4071-4084 (2006)
Three-step assembly of 4-aminotetrahydropyran-2-ones from isoxazoline-2-oxides.
Naumova AS, et al.
Royal Society of Chemistry Advances, 4(24), 12467-12475 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service