Skip to Content
Merck
All Photos(3)

Documents

51491

Sigma-Aldrich

Dibutyl sulfide

≥98.0% (GC)

Synonym(s):

1,1′-Thiobisbutane, Butyl sulfide, NSC 8460

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3S(CH2)3CH3
CAS Number:
Molecular Weight:
146.29
Beilstein:
1732829
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.07 (vs air)

vapor pressure

5.17 mmHg ( 37.7 °C)

Assay

≥98.0% (GC)

refractive index

n20/D 1.452 (lit.)
n20/D 1.453

bp

188-189 °C (lit.)

mp

−76 °C (lit.)

density

0.838 g/mL at 25 °C (lit.)

SMILES string

CCCCSCCCC

InChI

1S/C8H18S/c1-3-5-7-9-8-6-4-2/h3-8H2,1-2H3

InChI key

HTIRHQRTDBPHNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl sulfide is a volatile organic sulfur compound that is used as an organic building block in chemical synthesis. It is used to prepare dibutyl sulfoxide (DBSO) via oxidation reaction. DBSO is used as a valuable organic solvent for the extraction of zirconium, palladium, platinum, and gold.

Application

  • Environmental Odor Diagnostics: Dibutyl sulfide was utilized in developing a diagnostic method for complex odor issues in micro-polluted source water, highlighting its role in environmental monitoring and pollution control (Guo et al., 2021).
  • Surface Chemistry Investigations: The adsorption and manipulation of dibutyl sulfide on copper surfaces were studied, providing insights into its interactions at the molecular level (Jensen et al., 2007).
  • Biosensor Development for Chemical Warfare Detection: Dibutyl sulfide′s effects were explored in the context of stand-off tissue-based biosensors for detecting chemical warfare agents, utilizing photosynthetic fluorescence induction, it is crucial in constructing the inertial forces of launching or dropping into suspected danger zones and, with encrypted telecommunication data transmission (Sanders et al., 2001).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of irradiation intensity on rate and selectivity of photosensitized oxidation of dibutyl sulfide over C70 fullerene.
Arsentyev AV, et al.
High Energy Chemistry, 49(3), 167-172 (2015)
Dibutyl sulfide: synthesis, Pd, Pt extraction and separation studies.
Lu P, et al.
Chinese Journal of Inorganic Chemistry / Wu Ji Hua Xue Xue Bao, 24(4), 520-526 (2008)
Dithiolane-Directed Tandem Oxidation/1, 2-Benzyl Migration of Tetramic Acids under Ambient Conditions.
Li Y, et al.
Advanced Synthesis & Catalysis, 354(9), 1712-1716 (2012)
Revisiting the Zincke disulfide reaction for the post-polymerization functionalization of unsaturated polyolefins.
Dunst A, et al.
Polymer, 55(2), 5557-5560 (2014)
Adsorption, interaction, and manipulation of dibutyl sulfide on Cu {111}.
Jensen SC, et al.
ACS Nano, 1(1), 22-29 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service