Skip to Content
Merck
All Photos(2)

Key Documents

513040

Sigma-Aldrich

5,5′-Dimethyl-2,2′-dipyridyl

98%

Synonym(s):

5,5′-Dimethyl-2,2′-bipyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2
CAS Number:
Molecular Weight:
184.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

114-117 °C (lit.)

SMILES string

Cc1ccc(nc1)-c2ccc(C)cn2

InChI

1S/C12H12N2/c1-9-3-5-11(13-7-9)12-6-4-10(2)8-14-12/h3-8H,1-2H3

InChI key

PTRATZCAGVBFIQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and crystal structure determination of iron (III) hetero-ligand complexes containing 2, 2'-bipyridine, 5, 5'-dimethyl-2, 2'-bipyridine and chloride, [Fe(bipy)Cl4][bipy?H] and [Fe(dmbipy)2Cl2][FeCl4].
Amani V, et al.
Polyhedron, 26(15), 4257-4262 (2007)
Masaki Yoshida et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 36(1), 67-73 (2019-11-07)
The electrochemical and photophysical properties of a heteroleptic Cu(I) complex bearing an aliphatic α-diimine ligand, [Cu(dab)(xantphos)]+ (Cu-dab; dab = N,N'-diphenyl-2,3-dimethyl-1,4-diazabutadiene, xantphos = 4,5-bis(diphenylphosphino)-9,9-dimethylxanthene), were evaluated together with those of complexes [Cu(dmp)(xantphos)]+ (Cu-dmp; dmp = 2,9-dimethyl-1,10-phenanthroline), [Cu(dmbpy)(xantphos)]+ (Cu-dmbpy; dmbpy = 5,5'-dimethyl-2,2'-bipyridine)
Yuchen Wang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(66), 15170-15182 (2020-07-09)
In this study, we newly designed and synthesized a small library of ten structurally related C,N-cyclometalated ruthenium(II) complexes containing various pyridine-functionalized NHC ligand and chelating bipyridyl ligands (e.g., 2,2'-bipyridine, 5,5'-dimethyl-2,2'-bipyridine, and 1,10-phenanthroline (phen)). The complexes were well characterized by NMR
Muhammad Altaf et al.
Oncotarget, 8(1), 490-505 (2016-11-27)
We synthesized, characterized and tested in a panel of cancer cell lines, nine new bipyridine gold(III) dithiocarbamate-containing complexes. In vitro studies demonstrated that compounds 1, 2, 4, 5, 7 and 8 were the most cytotoxic in prostate, breast, ovarian cancer
Saikat Santra et al.
Dalton transactions (Cambridge, England : 2003), 46(39), 13300-13313 (2017-08-05)
Herein, a bis-amido tris-amine macrocycle and five bipyridine-based bidentate chelating ligands were investigated towards various divalent transition metal ion (Ni

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service