Skip to Content
Merck
All Photos(1)

Key Documents

511544

Sigma-Aldrich

5-Hydroxy-1-indanone

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O2
CAS Number:
Molecular Weight:
148.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

impurities

<3% acetone

mp

175 °C (dec.) (lit.)

functional group

ketone

SMILES string

Oc1ccc2C(=O)CCc2c1

InChI

1S/C9H8O2/c10-7-2-3-8-6(5-7)1-4-9(8)11/h2-3,5,10H,1,4H2

InChI key

ZRKQOVXGDIZYDS-UHFFFAOYSA-N

General description

5-Hydroxy-1-indanone is a 1-indanone derivative.

Application

5-Hydroxy-1-indanone may be used in the preparation of (5-hydroxy-indan-(1E)-ylidene)-acetic acid and 5-[2-(phenyl)ethoxy]-1-indanone. It may be used as a starting material in the multi-step synthesis of 5H-indeno[1,2-c]pyridazin-5-one analogs.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On the involvement of single-bond rotation in the primary photochemistry of photoactive yellow protein.
Stahl AD, et al.
Biophysical Journal, 101(5), 1184-1192 (2011)
Geometric and solvent effects on intramolecular phenolic hydrogen abstraction by carbonyl n,p* and p,p* triplets.
Lathioor EC and Leigh WJ.
Canadian Journal of Chemistry, 79(12), 1851-1863 (2001)
J Reniers et al.
European journal of medicinal chemistry, 46(12), 6104-6111 (2011-10-25)
Previous studies on 5H-indeno[1,2-c]pyridazin-5-one derivatives as inhibitors of MAO-B revealed that it was possible to increase the MAO-B inhibitory potency of 5H-indeno[1,2-c]pyridazin-5-ones by substituting the central heterocycle in the 3-position or C-8 with lipophilic groups which occupy the substrate cavity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service