Skip to Content
Merck
All Photos(1)

Key Documents

49914

Sigma-Aldrich

1-Benzyl-3-methylimidazolium chloride

≥97.0%

Synonym(s):

1-Methyl-3-benzylimidazolium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13ClN2
CAS Number:
Molecular Weight:
208.69
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0%

form

solid

functional group

phenyl

SMILES string

[Cl-].C[n+]1ccn(Cc2ccccc2)c1

InChI

1S/C11H13N2.ClH/c1-12-7-8-13(10-12)9-11-5-3-2-4-6-11;/h2-8,10H,9H2,1H3;1H/q+1;/p-1

InChI key

FCRZSGZCOGHOGF-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maleesha De Silva et al.
Journal of mass spectrometry : JMS, 55(9), e4518-e4518 (2020-06-25)
Ionic liquids are now used in applications ranging from chemical synthesis to spacecraft propulsion. With this comes the need to characterize new syntheses, identify environmental contamination, and determine eventual fate in terrestrial and space environments. This work investigates the effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service