Skip to Content
Merck
All Photos(1)

Key Documents

494348

Sigma-Aldrich

1-Bromo-4-methylpentane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CH(CH2)3Br
CAS Number:
Molecular Weight:
165.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.446 (lit.)

bp

146 °C (lit.)

density

1.134 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCCBr

InChI

1S/C6H13Br/c1-6(2)4-3-5-7/h6H,3-5H2,1-2H3

InChI key

XZKFBZOAIGFZSU-UHFFFAOYSA-N

General description

1-Bromo-4-methylpentane can be synthesized by treating 4-methyl-1-pentanol with phosphorous tribromide. The conformational analysis of its liquid-state and solid-state IR and Raman spectra and showed the presence of a mixture of PC,PH, and PH conformers.

Application

1-Bromo-4-methylpentane may be used in the synthesis of:
  • diethyl 2-(4′-methylpentyl)malonate
  • 1,3-bis(4-(isopentyloxy)phenyl)urea
  • 1,3-bis(4-(isopentyloxy)phenyl)thiourea
  • 13-methyl-1-[(tetrahydropyran-2-yl)oxy]tetradec-8-yne
  • 15-methyl-1-[(tetrahydropyran-2-yl)oxy]hexadec-10-yne
  • 3-[2-isohexyloxy-3-(hydroxymethyl]-5-phenyl-2-isohexyloxybenzyl alcohol

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria E Amato et al.
Molecules (Basel, Switzerland), 15(3), 1442-1452 (2010-03-26)
A novel chiral macrocyclic ligand incorporating a chiral salen moiety into a framework containing two biphenyl units was synthesized. Structural properties and conformational aspects of the free ligand and an UO2 complex were studied by using NMR spectroscopy in solution
E Takano et al.
The Journal of biological chemistry, 275(15), 11010-11016 (2001-02-07)
Early stationary phase culture supernatants of Streptomyces coelicolor A3(2) contained at least four small diffusible signaling molecules that could elicit precocious antibiotic synthesis in the producing strain. The compounds were not detected in exponentially growing cultures. One of these compounds
Néstor M Carballeira et al.
Chemistry and physics of lipids, 145(1), 37-44 (2006-11-28)
The first total syntheses for the (Z)-15-methyl-10-hexadecenoic acid and the (Z)-13-methyl-8-tetradecenoic acid were accomplished in seven steps and in 31-32% overall yields. The (trimethylsilyl)acetylene was the key reagent in both syntheses. It is proposed that the best synthetic strategy towards
B Reiter et al.
Journal of chemical ecology, 29(10), 2235-2252 (2003-12-20)
Gas chromatography, coupled gas chromatography-mass spectrometry (electron impact mode and chemical ionization with methane as reactant gas), gas chromatography-infrared spectroscopy, and derivatization techniques were used to identity 53 compounds in the interdigital secretion of the red hartebeest, Alcelaphus buselaphus caama.
Vibrational analysis of alkyl bromides: Part III. Branched-chain bromides: 1-bromo-3-methylbutane and 1-bromo-4-methylpentane.
Crowder GA and Jalilian MR.
Journal of Molecular Structure, 42, 71-76 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service