Skip to Content
Merck
All Photos(1)

Key Documents

478075

Sigma-Aldrich

2-Bromo-3-hydroxy-4-methoxybenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(OH)(OCH3)CHO
CAS Number:
Molecular Weight:
231.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

202-207 °C (lit.)

functional group

aldehyde
bromo

SMILES string

COc1ccc(C=O)c(Br)c1O

InChI

1S/C8H7BrO3/c1-12-6-3-2-5(4-10)7(9)8(6)11/h2-4,11H,1H3

InChI key

QPDFBPIHEDAUKK-UHFFFAOYSA-N

General description

2-Bromo-3-hydroxy-4-methoxybenzaldehyde, also known as 2-bromo-isovanillin, can be synthesized by the bromination of 3-hydroxy-4-methoxybenzaldehyde.

Application

2-Bromo-3-hydroxy-4-methoxybenzaldehyde (2-Bromo-isovanillin) may be used in the preparation of:
  • 2-hydroxy-3-methoxybenzaldehyde semicarbazone (HMBS)
  • 2-cyclopentyl-7-methoxy-1-benzofuran-4-carbaldehyde
  • 3-(benzyloxy)-2-bromo-4-methoxybenzaldehyde
It may also be used as a starting material in the total synthesis of the following natural products:
  • pareitropone
  • denbinobin
  • (±)-codeine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of pareitropone via radical anion coupling.
Hong SK, et al.
Organic Letters, 12(17), 3954-3956 (2010)
A concise synthesis of denbinobin.
Wang YC, et al.
Tetrahedron Letters, 46(47), 8103-8104 (2005)
Synthesis and preliminary biological evaluation of novel taspine derivatives as anticancer agents.
Zhang J, et al.
European Journal of Medicinal Chemistry, 45(7), 2798-2805 (2010)
Chemoselective Zinc/HCl Reduction of Halogenated β-Nitrostyrenes: Synthesis of Halogenated Dopamine Analogues.
Maresh JJ, et al.
Synlett, 25, 2891-2894 (2014)
Concise syntheses of (-)-galanthamine and (?)-codeine via intramolecular alkylation of a phenol derivative.
Magnus P, et al.
Journal of the American Chemical Society, 131(44), 16045-16047 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service