471607
2-Bromopropiophenone
95%
Synonym(s):
α-Bromopropiophenone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5COCHBrCH3
CAS Number:
Molecular Weight:
213.07
Beilstein:
508550
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
refractive index
n20/D 1.571 (lit.)
bp
245-250 °C (lit.)
density
1.4 g/mL at 25 °C (lit.)
functional group
bromo
ketone
phenyl
storage temp.
2-8°C
SMILES string
CC(Br)C(=O)c1ccccc1
InChI
1S/C9H9BrO/c1-7(10)9(11)8-5-3-2-4-6-8/h2-7H,1H3
InChI key
WPDWOCRJBPXJFM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
2-Bromopropiophenone (α-Bromopropiophenone) is an α-bromoketone. It participates in the manufacturing process of 1-acetamido-3-methyl-2-phenylimidazo[1,2-a]pyridinium bromide.
Application
2-Bromopropiophenone (α-Bromopropiophenone) may be used for the synthesis of 2-phenylmorpholinols. It may also be used in the synthesis of N-phenacylammonium salts [N-(α-benzoylbenzyl)-, N-(1-benzoylethyl)-, N-phenacyl-, pyrazinium, 3-bromoquinolinium, benzothiazolium, or p-cyanopyridinium hexafluoroantimonates].
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Photocationic and radical polymerizations by novel N-phenacylammonium salts.
Takahashi E, et al.
Journal of Applied Polymer Science, 91(6), 3470-3476 (2004)
William Andrew Publishing
Pharmaceutical Manufacturing Encyclopedia, 1-4, 1552-1552 null
Synthesis, stereochemistry and anti-tetrabenazine activity of bicyclo analogues of 2-phenylmorpholines.
Boswan EG, et al.
Journal of Heterocyclic Chemistry, 34(6), 1813-1820 (1997)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service