Skip to Content
Merck
All Photos(2)

Documents

461814

Sigma-Aldrich

5-Bromosalicylic acid

technical grade, 90%

Synonym(s):

5-Bromo-2-hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3-2-(OH)CO2H
CAS Number:
Molecular Weight:
217.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

solid

mp

159-162 °C (lit.)

SMILES string

OC(=O)c1cc(Br)ccc1O

InChI

1S/C7H5BrO3/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,9H,(H,10,11)

InChI key

IEJOONSLOGAXNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Bromosalicylic acid is a salicylic acid derivative. Its bonding to viscose fabric can impart antibacterial properties to the fabric. It has been identified as one of the brominated disinfection byproduct (Br-DBP) formed during the chlorination of saline sewage effluents.

Application

5-Bromosalicylic acid may be employed as a starting reagent for the synthesis of honokiol, a biphenyl-type neolignan.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongrui Cheng et al.
Nanoscale, 12(17), 9471-9480 (2020-04-30)
Highly flexible and stable plasmonic nanopaper comprised of silver nanocubes and cellulose nanofibres was fabricated through a self-assembly-assisted vacuum filtration method. It shows significant enhancement of the fluorescence emission with an enhancement factor of 3.6 and Raman scattering with an
Jian Zhu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 236, 118343-118343 (2020-04-18)
Au@AuAg yolk-shell nanorods with tunable and uniform interior gap were synthesized through galvanic replacement reaction, where Au@Ag core-shell nanorods served as sacrificial templates and HAuCl4 solution served as reductant. The effects of HAuCl4, Ag shell thickness and aspect ratio (AR)
Álvaro Cárcamo-Martínez et al.
Nanomaterials (Basel, Switzerland), 10(3) (2020-03-27)
Current strategies for the treatment of superficial non-melanoma skin cancer (NMSC) lesions include topical imoquimod, 5-fluorouracil, and photodynamic therapy. Although these treatments are effective, burning pain, blistering, and dermatitis have been reported as frequent side effects, making these therapies far
Yi Wang et al.
Analytica chimica acta, 1002, 97-104 (2018-01-08)
Seed-mediated growth has been employed as a simple and powerful means to the shape-controlled synthesis of metal nanocrystals. In this work, we apply the principle of seed-mediated growth in analytical chemistry, and achieve improved sensitivity due to the low energy
Tomoyuki Esumi et al.
Bioorganic & medicinal chemistry letters, 14(10), 2621-2625 (2004-04-28)
Honokiol, a biphenyl-type neolignan, which shows the remarkable neurotrophic effect in primary cultured rat cortical neurons, has been effectively synthesized in 21% yield over 14 steps starting from 5-bromosalicylic acid and p-hydroxybenzoic acid by utilizing Pd-catalyzed Suzuki-Miyaura coupling reaction as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service