Skip to Content
Merck
All Photos(2)

Key Documents

456640

Sigma-Aldrich

Poly(oxy-1,4-phenyleneoxy-1,4-phenylenecarbonyl-1,4-phenylene)

Synonym(s):

PEEK, Polyether ether ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(OC6H4OC6H4COC6H4)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

pellets

hardness

126 (Rockwell R, ASTM D 785)

mp

322 °C

transition temp

Tm (DSC) 322 °C (onset)

solubility

all common solvents: insoluble
concentrated sulfuric acid: soluble

density

1.3 g/mL at 25 °C (lit.)

SMILES string

Oc1ccc(O)cc1.Oc2ccc(cc2)C(=O)c3ccc(O)cc3

InChI

1S/C13H10O3.C6H6O2/c14-11-5-1-9(2-6-11)13(16)10-3-7-12(15)8-4-10;7-5-1-2-6(8)4-3-5/h1-8,14-15H;1-4,7-8H

InChI key

JHGYTDAAMWIDRN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(oxy-1,4-phenyleneoxy-1,4-phenylenecarbonyl-1,4-phenylene)(PEEK) is a semi-crystalline polymer with good thermo-mechanical properties and high chemical resistance which can be used as engineering plastics.

Application

PEEK can be sulfonated and coated on nanofiltration(NF) membranes which can be used in the fabrication of antifouling based NF membranes. Sulfonated PEEK (up to 50-70 degree of sulfonation(DS)) can be used in the formation of proton exchange membranes(PEMs) for fuel cells based applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of sulfonated polyether-etherketone (SPEEK) and composite membranes on the proton exchange membrane fuel cell (PEMFC) performance.
Sengul E, et al.
International Journal of Hydrogen Energy, 34(10), 4645-4652 (2009)
Sulfonated poly (ether ether ketone) membranes for direct methanol fuel cell.
Li L, et al.
Journal of Membrane Science , 226(1-2), 159-167 (2003)
Synthesis and characterization of sulfonated poly (ether ether ketone) for proton exchange membranes.
Xing P, et al.
Journal of Membrane Science , 229(1-2), 95-106 (2004)
Glass transition and melting behavior of poly (oxy-1, 4-phenyleneoxy-1, 4-phenylenecarbonyl-1, 4-phenylene)(PEEK).
Cheng SZD, et al.
Macromolecules, 19(7), 1868-1876 (1986)
Proton conducting composite membranes from polyether ether ketone and heteropolyacids for fuel cell applications.
Zaidi SMJ, et al.
Journal of Membrane Science , 173(1), 17-34 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service