Skip to Content
Merck
All Photos(1)

Key Documents

452807

Sigma-Aldrich

Trichloro(3,3,3-trifluoropropyl)silane

97%

Synonym(s):

3,3,3-Trifluoropropyl-trichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH2CH2SiCl3
CAS Number:
Molecular Weight:
231.50
Beilstein:
1748876
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.386 (lit.)

bp

114 °C (lit.)

density

1.419 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

FC(F)(F)CC[Si](Cl)(Cl)Cl

InChI

1S/C3H4Cl3F3Si/c4-10(5,6)2-1-3(7,8)9/h1-2H2

InChI key

WEUBQNJHVBMUMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trichloro(3,3,3-trifluoropropyl)silane can be used as a precursor for the preparation of:
  • Fluorinated polyhedral oligomeric silsesquioxane (F-POSS) by base-catalyzed condensation.        
  • 3,3,3-Trifluoropropyltrimethoxysilane by alcoholysis reaction with methanol.

It can also be used as a silanizing agent to coat the inner surface of pipettes in order to investigate the electrical charging of droplets discharged from the micropipette.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.7 °F - closed cup - (own results)

Flash Point(C)

45.4 °C - closed cup - (own results)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fabrication of Carbon Nanotube High-Frequency Nanoelectronic Biosensor for Sensing in High Ionic Strength Solutions
Journal of Visualized Experiments, 77, e50438-e50438 (2013)
Kyung Won Ro et al.
Electrophoresis, 24(18), 3253-3259 (2003-10-01)
Capillary electrochromatography (CEC) and preconcentration of neutral compounds have been realized on poly(dimethylsiloxane) (PDMS) microchips. The channels are coated with polyelectrolyte multilayers to avoid absorption of hydrophobic analytes into PDMS. The structures of a microchip include an injector and a
Dongwhi Choi et al.
Scientific reports, 3, 2037-2037 (2013-06-21)
We report that a droplet dispensed from a micropipette almost always has a considerable electrical charge of a magnitude dependent on the constituents of the droplet, on atmospheric humidity and on the coating material of pipette tip. We show that
Scott T Iacono et al.
Chemical communications (Cambridge, England), (47), 4992-4994 (2007-12-01)
New fluorinated polyhedral oligomeric silsesquioxane (F-POSS) structures possessing a high degree of hydrophobicity have been prepared via a facile corner-capping methodology.
Yu-Chih Chen et al.
Integrative biology : quantitative biosciences from nano to macro, 11(9), 353-361 (2019-12-11)
Considerable evidence suggests that breast cancer development and metastasis are driven by cancer stem-like cells (CSCs). Due to their unique role in tumor initiation, the interaction between CSCs and stromal cells is especially critical. In this work, we developed a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service