Skip to Content
Merck

450421

4,4′-Cyclohexylidenebisphenol

98%

Synonym(s):

Bisphenol Z

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H10(C6H4OH)2
CAS Number:
Molecular Weight:
268.35
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12162002
EC Number:
212-677-1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

98%

mp

190-192 °C (lit.)

SMILES string

Oc1ccc(cc1)C2(CCCCC2)c3ccc(O)cc3

InChI

1S/C18H20O2/c19-16-8-4-14(5-9-16)18(12-2-1-3-13-18)15-6-10-17(20)11-7-15/h4-11,19-20H,1-3,12-13H2

InChI key

SDDLEVPIDBLVHC-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Tian et al.
Food chemistry, 326, 126942-126942 (2020-05-15)
A non-targeted screening method based on ultrasound-assisted extraction followed by high-performance liquid chromatography coupled with quadrupole time-of-flight mass spectrometry (HPLC-QTOF-MS) was developed to screen for the presence of plastic-related chemicals (PRCs) in different types of food (fish, chicken, canned tuna
Jie Zhang et al.
Analytica chimica acta, 1032, 107-113 (2018-08-26)
A fluorescence polarization (FP) assay based on estrogen receptor was developed for the determination of bisphenol compounds (BPs). The human estrogen receptor α ligand binding domain (hERα-LBD) and coumestrol were employed as recognition element and fluorescent probe, respectively. Competitive displacement
Yi Chen et al.
Environmental pollution (Barking, Essex : 1987), 238, 299-305 (2018-03-25)
Bisphenols and triclosan (TCS) are widely used in consumer products. However, knowledge on human exposure to these anthropogenic chemicals has remained limited in China, especially for children. In this study, concentrations of seven bisphenols and TCS were determined in 283
Henry J Eichman et al.
Rapid communications in mass spectrometry : RCM, 31(20), 1773-1778 (2017-08-03)
Human health concerns have arisen for common bisphenols (BPA and BPS) as well as new structural analogs. While native sulfone bisphenols and sulfone-derivatized bisphenols are amenable to electrospray ionization (ESI), alternative ionization methods such as atmospheric pressure chemical ionization (APCI)
Marie-Katherin Zühlke et al.
Applied microbiology and biotechnology, 104(8), 3569-3583 (2020-03-04)
Comparative analyses determined the relationship between the structure of bisphenol A (BPA) as well as of seven bisphenol analogues (bisphenol B (BPB), bisphenol C (BPC), bisphenol E (BPE), bisphenol F (BPF), bisphenol Z (BPZ), bisphenol AP (BPAP), bisphenol PH (BPPH))

Global Trade Item Number

SKUGTIN
450421-25G04061833598412

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service