Skip to Content
Merck
All Photos(1)

Documents

445401

Sigma-Aldrich

Hexadecyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15NCO
CAS Number:
Molecular Weight:
267.45
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.448 (lit.)

bp

186-188 °C/14 mmHg (lit.)

density

0.861 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCN=C=O

InChI

1S/C17H33NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-18-17-19/h2-16H2,1H3

InChI key

GFLXBRUGMACJLQ-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongliang Liu et al.
Rapid communications in mass spectrometry : RCM, 22(13), 2109-2114 (2008-06-05)
Electron impact mass spectra of four long-chain isocyanates, lauryl isocyanate, tetradecyl isocyanate, hexadecyl isocyanate and octadecyl isocyanate, were obtained with a GCT high-resolution time-of-flight mass spectrometer. The four isocyanates studied gave a common base peak of m/z 99, which suggested
Joris Sprakel et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 77(6 Pt 1), 061502-061502 (2008-07-23)
We discuss the thermal motion of colloidal particles in transient polymer networks. For particles that are physically bound to the surrounding chains, light-scattering experiments reveal that the submillisecond dynamics changes from diffusive to Rouse-like upon crossing the network formation threshold.
Amrita Pal et al.
Langmuir : the ACS journal of surfaces and colloids, 29(7), 2120-2127 (2013-01-25)
A series of L-cysteine-derived double hydrocarbon chain amphiphilic gelators L-(3-alkyl-carbamoylsulfanyl)-2-(3-alkylurido)propionic acid with different hydrocarbon chain lengths (C6-C16) was designed and synthesized. These gelators efficiently gelate only aromatic solvents. The gelation ability increased with the increase of chain length up to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service