Skip to Content
Merck
All Photos(1)

Key Documents

445274

Sigma-Aldrich

Imino-tris(dimethylamino)phosphorane

97%

Synonym(s):

N,N,N′,N′,N″,N″-Hexamethylphosphorimidic triamide, Phosphazene base P1-H

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]3P=NH
CAS Number:
Molecular Weight:
178.22
Beilstein:
1933051
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.476 (lit.)

density

0.979 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CN(C)P(=N)(N(C)C)N(C)C

InChI

1S/C6H19N4P/c1-8(2)11(7,9(3)4)10(5)6/h7H,1-6H3

InChI key

GKTNLYAAZKKMTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Imino-tris(dimethylamino)phosphorane is a base that can be used:
  • As a reagent in the deboronation of ortho and meta carboranes.
  • As an organocatalyst in the synthesis of cyclic organic carbonates using glycerol under solvent-free conditions.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvent-free organocatalytic preparation of cyclic organic carbonates under scalable continuous flow conditions
Wang Z, et al.
Reaction Chemistry & Engineering, 4(1), 17-26 (2019)
Studies of icosahedral carboranes with Imino-tris(dimethylamino)phosphorane, HNP(NMe2)3
Contemporary Boron Chemistry (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service