Recommended Products
Assay
95%
form
solid
impurities
3% xylene
bp
160-164 °C/3 mmHg (lit.)
mp
57-61 °C (lit.)
functional group
hydroxyl
imide
SMILES string
OCCN1C(=O)CCC1=O
InChI
1S/C6H9NO3/c8-4-3-7-5(9)1-2-6(7)10/h8H,1-4H2
InChI key
TWYIPMITVXPNEM-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A new synthesis and crystal structure of n-(2-hydroxyethyl) succinimide.
Tetrahedron, 44(1), 261-265 (1988)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service