Skip to Content
Merck
All Photos(1)

Key Documents

435503

Sigma-Aldrich

Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol)

average Mn ~3,300

Synonym(s):

Pluronic® 31R1, PPG-PEG-PPG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

viscous liquid

mol wt

average Mn ~3,300

composition

PEG, 10 wt. %

refractive index

n20/D 1.454

surface tension

34 dyn/cm, 25 °C, 0.1 wt. % in H2O

viscosity

660 cP(25 °C, Brookfield)(lit.)

transition temp

softening point −25 °C
cloud point 25 °C (1 wt. % aqueous solution)

density

1.018 g/mL at 25 °C

HLB

2.0 - 7.0

SMILES string

O2C(C2)C.O1CC1

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

General description

Surface activity of the polymer is dependent on the EO/PO ratio. PPG-PEG-PPG exhibits lower foaming, greater defoaming and reduced gelling tendencies relative to Pluronic® surfactants. Terminal secondary hydroxyl groups of PPG-PEG-PPG have lower reactivity and acidity than the primary hydroxyl groups in Pluronic surfactants.

Application

Hard and soft surface cleaners, defoamers in coatings and water treatment. Lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.
PPG-PEG-PPG Pluronic® 31R1 can be used as surface cleaners (soft and hard), defoamers in coatings, in water treatment, lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.

Features and Benefits

Surface activity is dependent on the EO/PO ratio. Lower foaming, greater defoaming and reduced gelling tendencies relative to Pluronic® surfactants. Terminal secondary hydroxyl groups have lower reactivity and acidity than the primary hydroxyl groups in Pluronic® surfactants.

Legal Information

Pluronic is a registered trademark of BASF

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianan Shen et al.
Biomaterials, 34(5), 1581-1590 (2012-11-14)
The metastasis of breast cancer is the leading cause of cancer death in women, and the lung is a common location of a secondary tumor that has metastasized from the primary source tumor. In this work, an attempt to simultaneously
Irshad A Wani et al.
Colloids and surfaces. B, Biointerfaces, 101, 243-250 (2012-09-27)
Silver nanoparticles have been synthesized in the inverse microemulsions formed using three different surfactants viz., cetyl-trimethyl ammonium bromide (CTAB), Tergitol and Triton X-100. We have done a systematic study of the effect of the surfactants on the particle size and
Wendell J Lu et al.
Lipids, 47(6), 571-580 (2012-02-03)
Glucagon-like peptide-1 (GLP-1) and glucose-dependent insulinotropic polypeptide (GIP) are incretins produced in the intestine that play a central role in glucose metabolism and insulin secretion. Circulating concentrations of GLP-1 and GIP are low and can be difficult to assay in
Tsz Chung Lai et al.
Colloids and surfaces. B, Biointerfaces, 99, 27-37 (2011-10-18)
Polyplex particles formed with plasmid DNA (pDNA) and Pluronic P85-block-poly{N-[N-(2-aminoethyl)-2-aminoethyl]aspartamide} (P85-b-P[Asp(DET)]) demonstrated highly effective transfection ability compared to PEG-based block cationomer, PEG-b-P[Asp(DET)]. Ternary polyplexes comprising PEG-b-P[Asp(DET)], poly(ethylene oxide)-b-poly(propylene oxide)-b-poly(ethylene oxide)-b-P[Asp(DET)] (P(EPE)-b-P[Asp(DET)]) used as an analog of P85-b-P[Asp(DET)], and pDNA were
Houjian Gong et al.
Langmuir : the ACS journal of surfaces and colloids, 28(38), 13590-13600 (2012-09-08)
The block polyethers PEO-PPO-ph-PPO-PEO (BPE) and PPO-PEO-ph-PEO-PPO (BEP) are synthesized by anionic polymerization using bisphenol A as initiator. Compared with Pluronic P123, the aggregation behaviors of BPE and BEP at an air/water interface are investigated by the surface tension and

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service