Skip to Content
Merck
All Photos(3)

Key Documents

430943

Sigma-Aldrich

Poly(tetrafluoroethylene)

powder (free-flowing), ≤12 μm particle size

Synonym(s):

Fluorinated polymer, Fluoropolymer, PTFE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF2CF2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.25

form

powder (free-flowing)

Quality Level

particle size

≤12 μm

transition temp

Tm 321 °C

density

2.15 g/mL at 25 °C

SMILES string

F\C(F)=C(\F)F

InChI

1S/C2F4/c3-1(4)2(5)6

InChI key

BFKJFAAPBSQJPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Inert lubricant.

Physical properties

Low coefficient of friction, low critical surface energy and excellent antistick properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Yang et al.
The Journal of thoracic and cardiovascular surgery, 150(1), 152-157 (2015-03-15)
Robotically assisted cardiac surgery is an alternative to conventional, open-chest surgery. Although studies have been done on the clinical effect, morbidity, and mortality of robotically assisted atrial myxoma excision, few have addressed surgical outcomes, such as pain, quality of life
Marcel Ander et al.
PLoS biology, 13(8), e1002213-e1002213 (2015-08-14)
Repair of DNA breaks by single-strand annealing (SSA) is a major mechanism for the maintenance of genomic integrity. SSA is promoted by proteins (single-strand-annealing proteins [SSAPs]), such as eukaryotic RAD52 and λ phage Redβ. These proteins use a short single-stranded
Jianli Zou et al.
Nature communications, 5, 5254-5254 (2014-10-17)
Despite recent progress in preparing numerous types of nanosheets, it remains a difficult challenge to assemble the tiny building blocks into functional macroscale architectures suitable for practical applications. Here we introduce a diffusion driven layer-by-layer assembly process and demonstrate its
Tobias Fink et al.
Anesthesiology, 122(1), 117-126 (2014-08-30)
Multicapillary column ion-mobility spectrometry (MCC-IMS) may identify volatile components in exhaled gas. The authors therefore used MCC-IMS to evaluate exhaled gas in a rat model of sepsis, inflammation, and hemorrhagic shock. Male Sprague-Dawley rats were anesthetized and ventilated via tracheostomy
Seung Hee Jeong et al.
Scientific reports, 5, 8419-8419 (2015-02-13)
Stretchable electronics offers unsurpassed mechanical compliance on complex or soft surfaces like the human skin and organs. To fully exploit this great advantage, an autonomous system with a self-powered energy source has been sought for. Here, we present a new

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service