430862
Poly(ethylene-co-glycidyl methacrylate)
pellets, melt index 5 g/10 min (190°C/2.16kg)
Synonym(s):
2-Propenoic acid, 2-methyl-, oxiranylmethyl ester, polymer with ethene, Ethylene-glycidyl methacrylate polymer, Glycidyl methacrylate-ethylene copolymer
About This Item
Recommended Products
form
pellets
melt index
5 g/10 min (190°C/2.16kg)
composition
glycidyl methacrylate, 8 wt. %
hardness
92 (Shore A, ASTM D 2240)
mp
99 °C (lit.)
transition temp
softening point 87 °C (Vicat, ASTM D 1525-1kg)
density
0.94 g/mL at 25 °C (lit.)
SMILES string
O1C(C1)COC(=O)C(=C)C.C=C
InChI
1S/C7H10O3.C2H4/c1-5(2)7(8)10-4-6-3-9-6;1-2/h6H,1,3-4H2,2H3;1-2H2
InChI key
CONHAJWVOAJZGC-UHFFFAOYSA-N
Related Categories
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service