Skip to Content
Merck
All Photos(2)

Key Documents

427691

Sigma-Aldrich

3-Iodobenzyl bromide

95%

Synonym(s):

α-Bromo-m-iodotoluene, 1-(Bromomethyl)-3-iodobenzene, 3-(Bromomethyl)iodobenzene, m-Iodobenzylbromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4CH2Br
CAS Number:
Molecular Weight:
296.93
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

mp

46-51 °C (lit.)

functional group

bromo
iodo

SMILES string

BrCc1cccc(I)c1

InChI

1S/C7H6BrI/c8-5-6-2-1-3-7(9)4-6/h1-4H,5H2

InChI key

BACZSVQZBSCWIG-UHFFFAOYSA-N

General description

3-Iodobenzyl bromide (3-IBBr, m-iodobenzyl bromide) is a meta-isomer of iodobenzyl bromide. It can be synthesized by the bromination of m-iodotoluene.

Application

3-Iodobenzyl bromide (3-IBBr, m-iodobenzyl bromide) was used as a derivatization reagent for the extraction and purification of thiouracil (TU).
It may be used in the synthesis of the following:
  • meta-substituted phenylalanine derivatives
  • N6-substituted aristeromycin derivative
  • (N)-methanocarba-N6-(3-iodobenzyl)adenosine

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of some sulfonyl halides with trypsin.
Kuranova IP and Konareva NV.
Bulletin of the Academy of Sciences of the USSR, Division of chemical science, 22(10), 2251-2254 (1973)
Julie A L Kiebooms et al.
Applied and environmental microbiology, 80(23), 7433-7442 (2014-09-28)
In recent years, the frequent detection of the banned thyreostat thiouracil (TU) in livestock urine has been related to endogenous TU formation following digestion of glucosinolate-rich Brassicaceae crops. Recently, it was demonstrated that, upon in vitro digestion of Brassicaceae, fecal
Phase Transfer Catalyst (PTC) Catalyzed Alkylations of Glycinamides for Asymmetric Syntheses of alpha-Amino Acid Derivatives.
Park YS, et al.
Bull. Korean Chem. Soc., 22(9), 958-962 (2001)
Barbara Woźniak et al.
Journal of veterinary research, 62(4), 511-517 (2019-02-08)
In the European Union, the use of thyreostatic drugs for fattening slaughter animals has been banned since 1981 under Council Directive 81/602/EEC. For protection of consumer health against unwanted residues and in compliance with Directive 96/23, each EU country must
K A Jacobson et al.
Journal of medicinal chemistry, 43(11), 2196-2203 (2000-06-08)
Adenosine receptor agonists have cardioprotective, cerebroprotective, and antiinflammatory properties. We report that a carbocyclic modification of the ribose moiety incorporating ring constraints is a general approach for the design of A(1) and A(3) receptor agonists having favorable pharmacodynamic properties. While

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service