426261
Iodoethane-1-13C
99 atom % 13C, 99% (CP), contains copper as stabilizer
Synonym(s):
Ethyl-1-13C iodide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH313CH2I
CAS Number:
Molecular Weight:
156.96
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
isotopic purity
99 atom % 13C
Assay
99% (CP)
contains
copper as stabilizer
refractive index
n20/D 1.512 (lit.)
bp
69-73 °C (lit.)
mp
-108 °C (lit.)
density
1.962 g/mL at 25 °C
mass shift
M+1
storage temp.
2-8°C
SMILES string
C[13CH2]I
InChI
1S/C2H5I/c1-2-3/h2H2,1H3/i2+1
InChI key
HVTICUPFWKNHNG-VQEHIDDOSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Muta. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
161.6 °F - closed cup
Flash Point(C)
72.00 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service