Skip to Content
Merck
All Photos(1)

Key Documents

422320

Sigma-Aldrich

4-Bromopiperidine hydrobromide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10BrN · HBr
CAS Number:
Molecular Weight:
244.96
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

199-202 °C (lit.)

SMILES string

Br[H].BrC1CCNCC1

InChI

1S/C5H10BrN.BrH/c6-5-1-3-7-4-2-5;/h5,7H,1-4H2;1H

InChI key

LVTIZXGNLIKUQZ-UHFFFAOYSA-N

General description

4-Bromopiperidine hydrobromide is a salt of halogenopiperidine. Synthesis of tritium labelled N-aminopiperidine using 4-bromopiperidine as precursor has been reported.

Application

4-Bromopiperidine hydrobromide may be used as a starting material in the synthesis of [2H] piperidine hydrochloride.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Balázs Schäfer et al.
Journal of labelled compounds & radiopharmaceuticals, 58(1), 7-13 (2015-01-24)
Cell membrane association of proteins can be achieved by the addition of lipid moieties to the polypeptide chain, and such lipid-modified proteins have important biological functions. A class of cell surface proteins contains a complex glycosylphosphatidylinositol (GPI) glycolipid at the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service