Skip to Content
Merck
All Photos(2)

Key Documents

419087

Sigma-Aldrich

Barium trifluoromethanesulfonate

98%

Synonym(s):

Barium triflate, Trifluoromethanesulfonic acid barium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)2Ba
CAS Number:
Molecular Weight:
435.47
Beilstein:
3729346
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
grade:
for analytical purposes

grade

for analytical purposes

Assay

98%

mp

>300 °C (lit.)

functional group

fluoro
triflate

SMILES string

[Ba++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/2CHF3O3S.Ba/c2*2-1(3,4)8(5,6)7;/h2*(H,5,6,7);/q;;+2/p-2

InChI key

DXJURUJRANOYMX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Ba(CF3SO3) can be utilized as a precursor to synthesize:
  • Alkali metal and silver trifluoromethanesulfonates.
  • Sodium trifluoromethanesulfonate (sodium triflate) by reacting with sodium sulfate. Sodium triflate finds application in organic synthesis as an efficient catalyst as well as a reactant in catalytic asymmetric Mannich-type reactions, and Diels-Alder reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium 1, 1, 1-Trifluoromethanesulfonate
Surya Prakash GK and Mathew T
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
Daniel Lundberg et al.
Physical chemistry chemical physics : PCCP, 20(21), 14525-14536 (2018-05-17)
The structures of the N,N-dimethylformamide (dmf), N,N-dimethylacetamide (dma), and N,N-dimethylpropionamide (dmp) solvated strontium and barium ions have been determined in solution using large angle X-ray scattering and EXAFS spectroscopy. The strontium ion has a mean coordination number (CN) between 6.2
Lingyong Jiang et al.
Organic & biomolecular chemistry, 10(40), 8102-8107 (2012-09-06)
A silver triflate-catalyzed tandem reaction of N'-(2-alkynylbenzylidene)hydrazide with pyridyne is presented. Different outcomes are obtained, depending on the pyridynes utilized in the transformation.
Atsushi Kinbara et al.
The Journal of organic chemistry, 77(20), 8999-9005 (2012-09-26)
A cationic rhodium(I) complex- or In(OTf)(3)-catalyzed synthesis of phenol derivatives from cyclohex-2-enone having an ethoxycarbonyl-substituted alkyne has been achieved. This reaction proceeds via enolization and an intramolecular Alder-Rickert reaction.
B V Subba Reddy et al.
Organic letters, 15(3), 464-467 (2013-01-18)
A novel method has been devised for the synthesis of 2,4,5-trisubstituted pyrrole derivatives through the coupling of α-diazoketones with β-enaminoketones and esters using 10 mol % of Cu(OTf)(2). A wide range of 2,3-disubstituted indole derivatives were also prepared from α-diazoketones

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service