413801
Zirconium acetate solution
in dilute acetic acid
Synonym(s):
Acetic acid, zirconium salt
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
liquid
composition
Zr, 15.0-17.0% gravimetric
reaction suitability
core: zirconium
concentration
in dilute acetic acid
density
1.279 g/mL at 25 °C
SMILES string
CC(=O)O[Zr](OC(C)=O)(OC(C)=O)OC(C)=O
InChI
1S/4C2H4O2.Zr/c4*1-2(3)4;/h4*1H3,(H,3,4);/q;;;;+4/p-4
InChI key
MFFVROSEPLMJAP-UHFFFAOYSA-J
Related Categories
Application
Zirconium acetate solution is a metallic salt solution that can be used for a variety of applications such as:
- a zirconium based precursor for the development of zirconia coatings
- a stabilizer for colloidal solutions, controlled growth of ice crystals, water proofing, and flame retardant treatments
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Fabrication and characterization of spherical zirconia particles for direct force measurement using the atomic force microscope
Hook MS, et al.
Langmuir, 15(19), 6220-6225 (1999)
Electrochemical performance of solid oxide fuel cells having electrolytes made by suspension and solution precursor plasma spraying
Marr M, et al.
Journal of Power Sources, 245, 398-405 (2014)
Ice-structuring mechanism for zirconium acetate
Deville S, et al.
Langmuir, 28(42), 14892-14898 (2012)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service