409405
Chlorotris(trimethylsilyl)silane
97%
Synonym(s):
1,1,1,3,3,3-Hexamethyl-2-(trimethylsilyl)-2-chlorotrisilane, 2-Chloro-1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane, Tris(trimethylsilyl)chlorosilane, Tris(trimethylsilyl)silyl chloride
About This Item
Recommended Products
Quality Level
Assay
97%
form
solid
mp
50-52 °C (lit.)
SMILES string
C[Si](C)(C)[Si](Cl)([Si](C)(C)C)[Si](C)(C)C
InChI
1S/C9H27ClSi4/c1-11(2,3)14(10,12(4,5)6)13(7,8)9/h1-9H3
InChI key
HCOROOJWVHFMRJ-UHFFFAOYSA-N
Related Categories
General description
Application
Used for:
- Insertion into a P-P bond
- Diastereoselective aldol and cascade reactions
- Preparation of hypersilyl substituted monophosphines and diphosphines
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3 - Water-react 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
87.8 °F - closed cup
Flash Point(C)
31 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service