Skip to Content
Merck
All Photos(3)

Key Documents

408212

Sigma-Aldrich

Sodium methacrylate

99%

Synonym(s):

methacrylic acid, sodium salt, Methacrylic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2Na
CAS Number:
Molecular Weight:
108.07
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

powder

mp

300 °C (lit.)

SMILES string

[Na+].CC(=C)C([O-])=O

InChI

1S/C4H6O2.Na/c1-3(2)4(5)6;/h1H2,2H3,(H,5,6);/q;+1/p-1

InChI key

SONHXMAHPHADTF-UHFFFAOYSA-M

Application

  • Natural rubber composites with high strength, modulus, water-resistance, and thermal stability, prepared with cellulose nanofibrils and sodium methacrylate: Describes the preparation and properties of natural rubber composites enhanced with sodium methacrylate (T Noguchi et al., 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianxiu Li et al.
Journal of chromatography. A, 1634, 461669-461669 (2020-11-10)
Polymer-grafted media have been a focus of recent development for ion exchange chromatography (IEC) because of their capacity and uptake kinetics that can lead to high dynamic capacity in protein purification. This work is devoted to developing novel cation exchangers
Mohamed Bouri et al.
Talanta, 99, 897-903 (2012-09-13)
A rapid and selective method for the extraction and determination of catecholamines (CLs) from urine samples has been successfully developed using a magnetic molecularly imprinted polymer (MMIP) as a sorbent material. The MMIP has been prepared using dopamine hydrochloride (DA)
Riteshkumar J Patel et al.
Journal of biomedical materials research. Part A, 100(8), 2054-2062 (2012-05-15)
Exudate analysis indicated that both hydroxamated methacrylic acid-based (HX-MAA) and methacrylic acid-based (MAA) beads without hydroxamation, implanted in an air pouch with gelatin discs, elicited similar biological responses, with both resulting in high cell counts and chemokine (MCP3) levels, relative
Gustaf D Olsson et al.
Journal of molecular recognition : JMR, 25(2), 69-73 (2012-02-01)
A series of molecular dynamics simulations of prepolymerization mixtures for phenylalanine anilide imprinted co-(ethylene glycol dimethacrylate-methacrylic acid) molecularly imprinted polymers have been employed to investigate the mechanistic basis for template selective recognition in these systems. This has provided new insights
Yantao Fan et al.
Lab on a chip, 12(22), 4724-4731 (2012-08-04)
Autapse is an unusual type of synapse generated by a neuron on itself. The ability to monitor axonal growth of single neurons and autapse formation in three-dimensions (3D) may provide fundamental information relating to many cellular processes, such as axonal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service