Skip to Content
Merck
All Photos(1)

Documents

407593

Sigma-Aldrich

Methyl 4-bromobenzoate

99%

Synonym(s):

1-Bromo-4-(methoxycarbonyl)benzene, 4-Bromobenzoic acid methyl ester, 4-Methoxycarbonyl-1-bromobenzene, 4-Methoxycarbonylphenyl bromide, Methyl p-bromobenzoate, p-Bromobenzoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CO2CH3
CAS Number:
Molecular Weight:
215.04
Beilstein:
2045132
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

77-81 °C (lit.)

SMILES string

COC(=O)c1ccc(Br)cc1

InChI

1S/C8H7BrO2/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5H,1H3

InChI key

CZNGTXVOZOWWKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 4-bromobenzoate is a para-substituted aryl bromide. Mol­ecules of methyl 4-bromobenzoate are almost planar. The compound is isostructural with methyl 4-iodo­benzoate. The Zeeman effect on the N.Q.R. (nuclear quadrupole resonance) of methyl 4-bromobenzoate on single crystals, has been investigated by the ′geometric method′.

Application

Methyl 4-bromobenzoate may be used in the following syntheses:
  • Three-carbon-bridged 5-substituted furo[2,3-d]pyrimidine and 6-substituted pyrrolo[2,3-d]pyrimidine analogs, employed as antifolates.
  • Methyl 4-tri-n-butylstannylbenzoate.
  • Methyl 4-(2-pyridyl)benzoate by cross coupling reaction with lithium 2-pyridyltriolborate.
  • Methyl 4-imidazo[1,2-a]pyridin-3-ylbenzoate by ligand-free Pd(OAc)2 catalyzed reaction with Imidazo[1,2-a]pyridine.
  • Methyl (E)-4-[3-(2-methoxycarbonylvinyl)-thiophen-2-yl]acrylate by reacting with methyl (E)-3-(thiophen-3-y)lacrylate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphine-free palladium-catalyzed direct arylation of imidazo [1,2-a] pyridines with aryl bromides at low catalyst loading.
Fu HY, et al.
The Journal of Organic Chemistry, 77(9), 4473-4478 (2012)
Aleem Gangjee et al.
Journal of medicinal chemistry, 47(27), 6893-6901 (2004-12-24)
Bridge homologation of the previously reported classical two-carbon-bridged antifolates, a 5-substituted 2,4-diaminofuro[2,3-d]pyrimidine (1) [which is a 6-regioisomer of LY231514 (Alimta)] and a 6-subsituted 2-amino-4-oxopyrrolo[2,3-d]pyrimidine, afforded the three-carbon-bridged antifolates analogues 4 and 5, with enhanced inhibitory activity against tumor cells in
Methyl 4-bromobenzoate.
Bolte M and Wissler J.
Acta Crystallographica Section E, Structure Reports Online, 62(3), o1192-o1193 (2006)
Synthesis of Lithium 2-Pyridyltriolborate and its Cross-Coupling Reaction with Aryl Halides.
Yamamoto Y, et al.
Organic Syntheses, 88, 79-86 (2011)
Zeeman effect on the nuclear quadrupole resonance of 81Br in single crystals of methyl 4-bromobenzoate, 4, 4'-dibromodiphenyl-ether and 4, 4'-dibromodiphenylsulphide at 77 K.
Ambrosetti R, et al.
Molecular Physics, 28(2), 551-558 (1974)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service