Skip to Content
Merck
All Photos(2)

Key Documents

402400

Sigma-Aldrich

2-[4-(Dibutylamino)-2-hydroxybenzoyl]benzoic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]2NC6H3(OH)COC6H4CO2H
CAS Number:
Molecular Weight:
369.45
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

190-193 °C (lit.)

SMILES string

CCCCN(CCCC)c1ccc(c(O)c1)C(=O)c2ccccc2C(O)=O

InChI

1S/C22H27NO4/c1-3-5-13-23(14-6-4-2)16-11-12-19(20(24)15-16)21(25)17-9-7-8-10-18(17)22(26)27/h7-12,15,24H,3-6,13-14H2,1-2H3,(H,26,27)

InChI key

QPNFUBAIQZJEPO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service