Skip to Content
Merck
All Photos(3)

Key Documents

401439

Sigma-Aldrich

Zinc perchlorate hexahydrate

Synonym(s):

Perchloric acid zinc hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn(ClO4)2·6H2O
CAS Number:
Molecular Weight:
372.38
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystals and lumps

form

crystals and lumps

Quality Level

reaction suitability

reagent type: oxidant

SMILES string

O.O.O.O.O.O.[Zn++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/2ClHO4.6H2O.Zn/c2*2-1(3,4)5;;;;;;;/h2*(H,2,3,4,5);6*1H2;/q;;;;;;;;+2/p-2

InChI key

PADPILQDYPIHQQ-UHFFFAOYSA-L

General description

Zinc perchlorate hexahydrate is a hexahydrate salt of zinc widely used as an oxidation reagent and catalyst in various organic transformations. It also finds application in the field of rechargeable batteries.

Application

Zinc perchlorate hexahydrate can be used as a catalyst:
  • For acylation of electron-deficient phenols, sterically hindered alcohols and amines.
  • For the synthesis of 2-hydroxy sulfides by the opening of epoxide rings with thiols.
  • For solvent‐free condensation of carboxylic acids with alcohols.
It can also be used as:
  • An electrolyte for aqueous Zinc batteries.
  • A precursor for the synthesis of highly fluorescent ZnSe quantumdots.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photo-assisted synthesis of highly fluorescent ZnSe(S) quantum dots in aqueous solution
Guo-Yu Lan, et al.
Journal of Materials Chemistry, 17, 2661-2666 (2007)
Highly Efficient Solvent-Free Condensation of Carboxylic Acids with Alcohols Catalysed by Zinc Perchlorate Hexahydrate, Zn(ClO4)2?6 H2O
G Bartoli, et al.
Advanced Synthesis & Catalysis, 347, 33-38 (2005)
Zinc perchlorate hexahydrate [Zn(ClO4)2?6H2O] as acylation catalyst for poor nucleophilic phenols, alcohols and amines: Scope and limitations
Shivani, et al.
J. Mol. Catal. A: Chem., 264, 208-213 (2007)
Zinc perchlorate hexahydrate as a new and highly efficient catalyst for synthesis of 2-hydroxysulfides by opening of epoxide rings with thiols under solvent-free conditions: Application for synthesis of the key intermediate of diltiazem
Shivani and Asit K. Chakraborti
J. Mol. Catal. A: Chem., 263, 137-142 (2007)
Ultrastable Zinc Anodes Enabled by Anti-Dehydration Ionic Liquid Polymer Electrolyte for Aqueous Zn Batteries
Jiaqi Huang, et al.
ACS Applied Materials & Interfaces, 13, 4008-4016 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service