Skip to Content
Merck
All Photos(1)

Key Documents

396532

Sigma-Aldrich

2,2,2-Trifluoroethanol-d3

≥99.5 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CD2OD
CAS Number:
Molecular Weight:
103.06
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

≥99.5 atom % D

Assay

≥99% (CP)

form

liquid

technique(s)

NMR: suitable

impurities

≤0.05% water
water

refractive index

n20/D 1.3 (lit.)

bp

77-80 °C (lit.)

mp

−44 °C (lit.)

density

1.415 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]OC([2H])([2H])C(F)(F)F

InChI

1S/C2H3F3O/c3-2(4,5)1-6/h6H,1H2/i1D2,6D

InChI key

RHQDFWAXVIIEBN-IDPMSXFZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2,2-Trifluoroethanol-d3 (TFEd3) is 2,2,2-trifluoroethanol in which hydrogen of the –OH group is substituted with deuterium. It is a commonly used 1H NMR and 13C NMR solvent.

Application

Recent uses include a gas-phase study of the silaformamide ion and an investigation of substituent effects in photosolvolysis.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication and characterization of electrospun polyamide 66 fibers crosslinked by gamma irradiation.
Kang HK, et al.
Macromolecular Research, 19(4), 364-369 (2011)
Chemistry Letters (Jpn), 2015-2015 (1994)
Helicity of [α] (404-451) and [β](394-445) tubulin C-terminal recombinant peptides.
Jimenez M, et al.
Protein Science, 8(04), 788-799 (1999)
Organometallics, 14, 2652-2652 (1995)
Sara La Manna et al.
Biochimie, 138, 106-115 (2017-05-10)
Interleukin-22 (IL-22) belongs to the family of IL-10 cytokines and is involved in a wide number of human diseases, including inflammatory disorders and cancer pathology. The ligand-receptor complex IL-22/IL-22R plays a key role in several pathways especially in the regulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service