Skip to Content
Merck
All Photos(1)

Key Documents

390828

Sigma-Aldrich

o-Eugenol

98%

Synonym(s):

2-Allyl-6-methoxyphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2C6H3(OCH3)OH
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.538 (lit.)

bp

119-121 °C/12 mmHg (lit.)

density

1.068 g/mL at 25 °C (lit.)

functional group

allyl

SMILES string

COc1cccc(CC=C)c1O

InChI

1S/C10H12O2/c1-3-5-8-6-4-7-9(12-2)10(8)11/h3-4,6-7,11H,1,5H2,2H3

InChI key

LREHGXOCZVBABG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1A

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Schranz et al.
Food chemistry, 232, 808-819 (2017-05-12)
Guaiacol and its derivatives are commonly found in nature and are known for their characteristic smoky, clove-like and vanilla-like smells. However, there is no systematic investigation of the smell properties of structurally related guaiacol derivatives. In order to establish a
Nurhayat Tabanca et al.
Molecules (Basel, Switzerland), 24(13) (2019-07-03)
Ceratitis capitata, the Mediterranean fruit fly, is one of the most serious agricultural pests worldwide responsible for significant reduction in fruit and vegetable yields. Eradication is expensive and often not feasible. Current control methods include the application of conventional insecticides
Adrian A Franke et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 80, 241-246 (2015-03-24)
Betel nut chewing causes cancer in humans, including strong associations with head and neck cancer in Guam. In the search for biomarkers of betel chewing we sought to identify chemicals specific for the 3 most commonly consumed betel preparations in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service