Skip to Content
Merck
All Photos(1)

Documents

386456

Sigma-Aldrich

Diethylamine

purified by redistillation, 99.5%

Synonym(s):

N-ethyl-ethanamine, DEA, Diethamine, Diethyl amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NH
CAS Number:
Molecular Weight:
73.14
Beilstein:
605268
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.5 (vs air)

vapor pressure

14.14 psi ( 55 °C)
180 mmHg ( 20 °C)
3.51 psi ( 20 °C)

Assay

99.5%

form

liquid

autoignition temp.

594 °F

purified by

redistillation

expl. lim.

10.1 %

technique(s)

ELISA: suitable

refractive index

n20/D 1.385 (lit.)

pH

13 (20 °C, 100 g/L)

bp

55 °C (lit.)

mp

−50 °C (lit.)

density

0.707 g/mL at 25 °C (lit.)

SMILES string

CCNCC

InChI

1S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3

InChI key

HPNMFZURTQLUMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethylamine can be used as a better template for the crystallization of the silicoaluminophosphate molecular sieves (SAPO-34) with high purity and crystallinity as compared to other templates. It can react with furfural and cyanoacetic ester to form bicyclo[3.2.1]octanes. It can also be used as an amino functionalized monomer which can interact with chitosan to synthesize hydrogel with potential applications in drug delivery systems.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of SAPO-34 templated by diethylamine: Crystallization process and Si distribution in the crystals
Liu G, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 114(1-3), 416-423 (2008)
A New Synthetic Approach to Functionalized Bicyclo [3.2.1] octanes
Ismiev A, et al.
Russ. J. Gen. Chem., 88(7), 1533-1536 (2018)
A simple strategy for robust preparation and characterisation of hydrogels derived from chitosan and amino functional monomers for biomedical applications
Khan F, et al.
Journal of Material Chemistry B: Materials for Biology and Medicine, 6(31), 5115-5129 (2018)
Rakesh Naduvile Veedu et al.
The journal of physical chemistry. A, 112(40), 9742-9750 (2008-09-16)
Five aryliminopropadienones 4a- d have been synthesized by flash vacuum thermolysis (FVT) by using two different precursors in each case. These compounds were deposited at 50 K at a pressure of ca. 10(-6) mbar together with three different nucleophiles, namely
Doron Yariv et al.
Colloids and surfaces. B, Biointerfaces, 78(2), 185-192 (2010-04-07)
In this paper we examined feasible correlations between the structure of different lyotropic mesophases and transdermal administration of three diclofenac derivatives with varying degrees of kosmotropic or chaotropic properties, solubilized within the mesophases. It was found that the most chaotropic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service