Skip to Content
Merck
All Photos(2)

Key Documents

381721

Sigma-Aldrich

Sodium pentafluoropropionate

98%

Synonym(s):

Pentafluoropropionic acid sodium salt, Perfluoropropionic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CF2COONa
CAS Number:
Molecular Weight:
186.01
Beilstein:
3636648
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

226-229 °C (dec.) (lit.)

functional group

fluoro

SMILES string

[Na+].[O-]C(=O)C(F)(F)C(F)(F)F

InChI

1S/C3HF5O2.Na/c4-2(5,1(9)10)3(6,7)8;/h(H,9,10);/q;+1/p-1

InChI key

SCWLIHXXYXFUFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium pentafluoropropionate is an aryl fluorinated building block.

Application

Sodium pentafluoropropionate may be used in the preparation of:
  • tributyltin pentafluoropropionate (Bu3SnO2CC2F5)
  • trimethyltin pentafluoropropionate
  • new o-fluoroalkyl-benzenesulfonamide precursors

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atmospheric pressure deposition of fluorine-doped SnO2 thin films from organotin fluorocarboxylate precursors.
Mahon MF, et al.
Applied Organometallic Chemistry, 19(5), 658-671 (2005)
New fluoro intermediates for herbicidal sulfonylureas.
Hamprecht G, et al.
Pestic. Sci., 55(5), 566-570 (1999)
K F Faull et al.
Journal of neuroscience research, 27(4), 533-540 (1990-12-01)
In experimental protocols with humans and non-human primates, cerebrospinal fluid (CSF) concentrations of 3-methoxy-4-hydroxyphenylethylene glycol (MHPG), the predominant end-product of norepinephrine metabolism in the mammalian central nervous system (CNS), have been widely used as an index of the rate of
T Krishnamurthy et al.
Journal of chromatography, 355(1), 253-264 (1986-03-14)
The pentafluoropropionyl, (PFP) and trifluoroacetyl (TFA) esters of several naturally occurring and synthetically modified simple trichothecenes were synthesized in nanogram amounts and characterized. Optimum conditions for the gas chromatographic (GC) separation of these derivatives and their analysis by negative ion
C T Mant et al.
Journal of chromatography. A, 1125(2), 211-219 (2006-07-04)
The present study set out to investigate whether observed relative hydrophilicity/hydrophobicity values of positively charged side-chains (with Lys and Arg as representative side-chains) or hydrophobic side-chains (with Ile as the representative side-chain) were context-dependent, i.e., did such measured values vary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service