Skip to Content
Merck
All Photos(1)

Key Documents

37870

Sigma-Aldrich

3-(2,4-Dihydroxyphenyl)propionic acid

≥95.0% (HPLC)

Synonym(s):

2,4-Dihydroxyhydrocinnamic acid, Hydroumbellic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O4
CAS Number:
Molecular Weight:
182.17
Beilstein:
2694769
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (HPLC)

form

solid

mp

158-162 °C

functional group

carboxylic acid

SMILES string

OC(=O)CCc1ccc(O)cc1O

InChI

1S/C9H10O4/c10-7-3-1-6(8(11)5-7)2-4-9(12)13/h1,3,5,10-11H,2,4H2,(H,12,13)

InChI key

HMCMTJPPXSGYJY-UHFFFAOYSA-N

General description

3-(2,4-Dihydroxyphenyl)propionic acid is a 3,4-dihydroxyphenylalanine (DOPA) analog. It is a building block for heterocyclic compounds.

Application

3-(2,4-Dihydroxyphenyl)propionic acid (DHPPA) may be used as substrate for the o-diphenolase in microplate assays of serum and hemocyte supernatants from QXR3 and wild type oysters for the quantitation of o-diphenolase activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new synthesis of psoralene [(author's transl)].
O Dann et al.
Archiv der Pharmazie, 308(2), 121-130 (1975-02-01)
B Morin et al.
The Biochemical journal, 330 ( Pt 3), 1059-1067 (1998-05-23)
A major product of hydroxy-radical addition to tyrosine is 3, 4-dihydroxyphenylalanine (DOPA) which has reducing properties. Protein-bound DOPA (PB-DOPA) has been shown to be a major component of the stable reducing species formed during protein oxidation under several conditions. The
Kathryn Newton et al.
Developmental and comparative immunology, 28(6), 565-569 (2004-06-05)
QX is a fatal disease in Sydney rock oysters (Saccostrea glomerata) that results from infection by the protistan parasite, Marteilia sydneyi. Since 1997, the New South Wales Fisheries Service has bred S. glomerata for resistance to QX disease. The current
Violetta Mohos et al.
Biomolecules, 10(3) (2020-03-12)
Flavonoids are abundant polyphenols in nature. They are extensively biotransformed in enterocytes and hepatocytes, where conjugated (methyl, sulfate, and glucuronide) metabolites are formed. However, bacterial microflora in the human intestines also metabolize flavonoids, resulting in the production of smaller phenolic
Eszter Fliszár-Nyúl et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 131, 110732-110732 (2020-09-18)
Polyphenolic compounds (including flavonoids, chalcones, phenolic acids, and furanocoumarins) represent a common part of our diet, but are also the active ingredients of several dietary supplements and/or medications. These compounds undergo extensive metabolism by human biotransformation enzymes and the microbial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service