Skip to Content
Merck
All Photos(1)

Documents

371963

Sigma-Aldrich

Hexafluoropropene, trimer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CF3C(F)=CF2]3
CAS Number:
Molecular Weight:
450.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

bp

110-115 °C (lit.)

density

1.83 g/mL at 25 °C (lit.)

SMILES string

FC(F)=C(F)C(F)(F)F.FC(F)=C(F)C(F)(F)F.FC(F)=C(F)C(F)(F)F

InChI

1S/3C3F6/c3*4-1(2(5)6)3(7,8)9

InChI key

VJRSWIKVCUMTFK-UHFFFAOYSA-N

General description

Spectra of three isomeric trimers of hexafluoropropene have been investigated. It reacts with primary amines, via indirect substitution of fluorine atoms, to form the corresponding enamines and enimines.

Application

Hexafluoropropene, trimer may be used in the preparation of photochromic spiroindolinonaphthoxazine derivative. It may be used in the preparation of 2-trifluoromethyl-3-heptadifluoroisopropyl-2-perfluoropentene-4-(N′,N′-dimethylamino propyl) imino(I).

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Fluoro-tertiary and Quaternary Ammonium Cationic Surfactants [J].
Jingfeng LZ, et al.
Dyestuffs and Coloration / Ran Liao Yu Ran Shai, 1, 009-009 (2006)
J de Rooij et al.
Nature, 396(6710), 474-477 (1998-12-16)
Rap1 is a small, Ras-like GTPase that was first identified as a protein that could suppress the oncogenic transformation of cells by Ras. Rap1 is activated by several extracellular stimuli and may be involved in cellular processes such as cell
D Findik et al.
Journal of cellular biochemistry, 57(1), 12-21 (1995-01-01)
In addition to a role for de novo protein synthesis in apoptosis we have previously shown that activation of a protein phosphatase or loss of activity of a kinase is also important in radiation-induced apoptosis in human cells [Baxter, and
C C Geilen et al.
FEBS letters, 309(3), 381-384 (1992-09-14)
In this study, we report that the potent and selective inhibitor of cyclic AMP-dependent protein kinase, N-[2-bromocinnamyl(amino)ethyl]-5-isoquinolinesulfonamide (H-89) interferes with the incorporation of choline into phosphatidylcholine in HeLa cells. Treatment of cells with 10 microM H-89 for 1 h decreases
Jost Leemhuis et al.
The Journal of pharmacology and experimental therapeutics, 300(3), 1000-1007 (2002-02-28)
The small GTPase RhoA can retract cell extensions by acting on two Rho kinases (ROCKs). Activated protein kinase A (PKA) inhibits RhoA and induces extensions. The isoquinoline H89 inhibits PKA and thus should prevent the inactivation of RhoA. In kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service