Skip to Content
Merck
All Photos(2)

Key Documents

349488

Sigma-Aldrich

Azidotrimethyltin(IV)

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SnN3
CAS Number:
Molecular Weight:
207.85
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

reaction type: click chemistry

mp

117-120 °C (lit.)

SMILES string

C[Sn](C)(C)N=[N+]=[N-]

InChI

1S/3CH3.N3.Sn/c;;;1-3-2;/h3*1H3;;/q;;;-1;+1

InChI key

OSJRGDBEYARHLX-UHFFFAOYSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service