Skip to Content
Merck
All Photos(2)

Documents

348104

Sigma-Aldrich

Triphenyltin hydroxide

Synonym(s):

Fentin hydroxide, Hydroxytriphenylstannane, NSC 113243

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3SnOH
CAS Number:
Molecular Weight:
367.03
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

mp

124-126 °C (lit.)

SMILES string

O[Sn](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/3C6H5.H2O.Sn/c3*1-2-4-6-5-3-1;;/h3*1-5H;1H2;/q;;;;+1/p-1

InChI key

BFWMWWXRWVJXSE-UHFFFAOYSA-M

Application

Reactant involved in transmetalation reactions with Ir clusters

Reactant involved in synthesis of:
  • (E)-(Nitrophenyl)propenoic acid organotin(IV) carboxylates with antitumor activity
  • Triorganotin chrysanthemumates for larvicidal studies
  • Polynuclear organotin(IV) carboxylates containing ferrocenyl moieties for antitumor investigation
  • Organotin(IV) aminophenylacrylate complexes
  • 4-Biphenylcarboxylic acid derivatives for in vitro cytotoxicity studies

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David I Shapiro-Ilan et al.
Environmental entomology, 40(1), 59-65 (2011-12-21)
Hypocreales fungi such as Beauveria bassiana (Balsamo) Vuillemin and Metarhizium brunneum Petch can be negatively affected by fungicides thereby reducing their biocontrol potential. In a previous study, we demonstrated enhanced fungicide resistance in B. bassiana through artificial selection. However, it
Eduardo C Oliveira-Filho et al.
Chemosphere, 81(2), 218-227 (2010-07-03)
Plant molluscicides have been regarded as possible alternatives to the costly and environmentally hazardous molluscicides currently available. This study was undertaken to compare the developmental toxicity of a plant molluscicide (Euphorbia milii latex, LAT) with that of three synthetic molluscicidal
S Tanimoto et al.
Molecular reproduction and development, 39(4), 409-414 (1994-12-01)
Flux of K+ and changes in intracellular Ca2+ in the sperm of salmonid fishes were measured with spectrophotometry, ion electrode, microscopic fluorometry, and radioisotope accumulation. Release of K+ occurred at the initiation of sperm motility which is induced by decrease
G E Walsh et al.
Ecotoxicology and environmental safety, 12(1), 95-100 (1986-08-01)
Effects of water-bourne toxicants on regeneration of arms by the brittle star, Ophioderma brevispina, are described. Regeneration was inhibited by 0.1 micrograms liter-1 bis(tri-n-butyltin)oxide and bis(triphenyltin)oxide. Both substances are known to act upon the nervous system, and it is suggested
Marcia Sarpa et al.
Regulatory toxicology and pharmacology : RTP, 49(1), 43-52 (2007-07-10)
Triphenyltin-hydroxide (TPTH) is used as agricultural fungicide in Brazil and elsewhere. This study was undertaken to evaluate the developmental toxicity of TPTH in mice. Swiss Webster mice were treated by gavage with TPTH (0, 3.75, 7.5, 15 and 30 mg/kg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service