Skip to Content
Merck
All Photos(2)

Key Documents

345172

Sigma-Aldrich

Tin(IV) acetate

Synonym(s):

Stannic Acetate, Tetraacetoxytin, Tin(4+) Diethanoate, Tin(4+) tetraacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sn(CH3CO2)4
CAS Number:
Molecular Weight:
354.89
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Sn, 31.4-35.5% gravimetric

reaction suitability

core: tin

mp

232-233 °C (lit.)

SMILES string

CC(=O)O[Sn](OC(C)=O)(OC(C)=O)OC(C)=O

InChI

1S/4C2H4O2.Sn/c4*1-2(3)4;/h4*1H3,(H,3,4);/q;;;;+4/p-4

InChI key

YJGJRYWNNHUESM-UHFFFAOYSA-J

General description

Tin(IV) acetate is a white crystalline solid widely used as a catalyst in organic transformations and a sol-gel precursor to synthesize nanomaterials.

Application

Tin(IV) acetate can be used:
  • As a reducing agent to synthesize silver nanoparticles for conductive inks for printed electronics.
  • As a precursor to prepare tin oxide nanopowder for gas sensors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miki Asanagi et al.
The Journal of toxicological sciences, 41(2), 207-215 (2016-03-11)
Organotin compounds, such as tributyltin (TBT), are well-known endocrine-disrupting chemicals (EDCs). We have recently reported that TBT induces growth arrest in the human embryonic carcinoma cell line NT2/D1 at nanomolar levels by inhibiting NAD(+)-dependent isocitrate dehydrogenase (NAD-IDH), which catalyzes the
Zhi-Yin Yang et al.
Molecules (Basel, Switzerland), 25(8) (2020-04-29)
Sn(II) binds to kaempferol (HKaem, 3,4',5,7-tetrahydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one) at the 3,4-site forming [Sn(II)(Kaem)2] complex in ethanol. DPPH• scavenging efficiency of HKaem is dramatically decreased by SnCl2 coordination due to formation of acid inhibiting deprotonation of HKaem as ligands and thus reduces the
Miika Mattinen et al.
Small (Weinheim an der Bergstrasse, Germany), 14(21), e1800547-e1800547 (2018-04-20)
Semiconducting 2D materials, such as SnS2 , hold immense potential for many applications ranging from electronics to catalysis. However, deposition of few-layer SnS2 films has remained a great challenge. Herein, continuous wafer-scale 2D SnS2 films with accurately controlled thickness (2
Comparative study of nanocrystalline SnO2 materials for gas sensor application: Thermal stability and catalytic activity
R.G. Pavelko, et al
Sensors and Actuators B, Chemical, 137, 637-643 (2009)
Seung Hyuk Lee et al.
Physical chemistry chemical physics : PCCP, 21(10), 5674-5678 (2019-02-26)
Plasmon-induced charge separation (PICS) by continuous electron injection from plasmonic compound nanoparticles to a semiconductor was achieved by using solid-state cells based on tin-doped indium oxide (ITO) nanoparticles with a short capping agent and a TiO2 film. The cells extended

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service