Skip to Content
Merck
All Photos(2)

Key Documents

330744

Sigma-Aldrich

Tetrapropylammonium perruthenate

97%

Synonym(s):

TPAP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)4NRuO4
CAS Number:
Molecular Weight:
351.43
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reagent type: oxidant

mp

~160 °C (dec.) (lit.)

SMILES string

[O-][Ru](=O)(=O)=O.CCC[N+](CCC)(CCC)CCC

InChI

1S/C12H28N.4O.Ru/c1-5-9-13(10-6-2,11-7-3)12-8-4;;;;;/h5-12H2,1-4H3;;;;;/q+1;;;;-1;

InChI key

NQSIKKSFBQCBSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrapropylammonium perruthenate is a mild oxidizing agent used for the oxidation of alcohols to corresponding carbonyl compounds. It is a non-volatile, air-stable, and readily soluble reagent, which can be used either stoichiometrically or catalytically with a suitable co-oxidant.

Application

Tetrapropylammonium perruthenate (TPAP) can be used as a catalyst:     
  • For the conversion of sulfides to sulfones by oxidation reaction.       
  • In the isomerization of allylic alcohols into the corresponding saturated carbonyl derivatives.      
  •  Along with N-methylmorpholine N-oxide (NMO) for the cleavage of glycol to carboxylic acids.

TPAP can also be used as an oxidizing reagent:
  • For the oxidation of benzyl alcohol to benzaldehyde and steroidal alcohols to corresponding ketones.
  • To convert N,N′-dihydroxyimidazolidines to nitronyl nitroxide free radicals.
  • To oxidize hydroxyl-substituted tri-n-butylammonium trifluoroborates to aldehydes and ketones without concomitant cleavage of the carbon-boron bond.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gary A Molander et al.
Journal of the American Chemical Society, 128(30), 9634-9635 (2006-07-27)
Potassium- and tetra-n-butylammonium organotrifluoroborates containing hydroxyl groups have been prepared and oxidized using several common oxidants. Aryl-, alkenyl-, and alkyltrifluoroborates containing both primary and secondary hydroxyl groups were oxidized in moderate to excellent yields with complete retention of the trifluoroborate
Tetrapropylammonium Perruthenate, Pr4N+RuO4 -, TPAP: A Catalytic Oxidant for Organic Synthesis
Steven V.L, et al.
Synthesis, 7, 639-666 (1994)
Aldrichimica Acta, 23, 13-13 (1990)
Tetrapropylammonium perruthenate catalyzed glycol cleavage to carboxylic (di) acids
Schmidt A-KC and Stark CBW
Organic Letters, 13(21), 5788-5791 (2011)
Synthesis, 639-639 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service