All Photos(3)
About This Item
Linear Formula:
H2C=CHC6H4SO3Na·xH2O
CAS Number:
Molecular Weight:
206.19 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
powder
Quality Level
SMILES string
[Na+].[H]O[H].[O-]S(=O)(=O)c1ccc(C=C)cc1
InChI
1S/C8H8O3S.Na.H2O/c1-2-7-3-5-8(6-4-7)12(9,10)11;;/h2-6H,1H2,(H,9,10,11);;1H2/q;+1;/p-1
InChI key
AATHLPHPRXGBAI-UHFFFAOYSA-M
General description
4-Styrenesulfonic acid sodium salt hydrate is a water soluble vinyl monomer. It can be polymerized to form copolymers/homopolymers with other vinyl monomers.
Application
4-Styrenesulfonic acid sodium salt hydrate (NaSS) modified membranes have been coated on proteins for lysozyme fractionation. NaSS can also be used in combination with potassium methacrylate to create ionic charges on the surface of the nanosilica for the fabrication of organic-inorganic hybrid nanoparticles. Silica nanoparticles can be grafted by radical polymerizing NaSS with 3-sulfopropylmethacrylate, which can be used in the preparation of proton-conducting membranes based nanoparticles (PBNPs) as components for fuel cells.
4-Styrenesulfonic acid sodium salt hydrate( NaSS, SSAS) was grafted on poly(ethersulfone) (PES) membrane to determine the fractionation of proteins with similar molecular weights and close pI values. NaSS based ion exchange membranes for membrane capacitive deionization (MCDI) have been synthesized. Copolymerization characteristics of sodium styrenesulfonate has been studied.c
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Preparation and properties of DMFC membranes from polymer-brush nanoparticles
Khabibullin A, et al.
Solid State Ionics, 288(6), 154-159 (2016)
Synthesis and electrical properties of NaSS-MAA-MMA cation exchange membranes for membrane capacitive deionization (MCDI)
Kwak NS, et al.
Desalination, 284, 138-146 (2012)
Copolymerization characteristics of sodium styrenesulfonate
Grabiel CE and Decker DL
Journal of Polymer Science, 59(168) (2003)
In situ synthesis of silica/polyacrylate nanocomposite particles simultaneously bearing carboxylate and sulfonate functionalities via soap-free seeded emulsion polymerization
Riazi H, et al.
Materials Chemistry and Physics, 207(6), 470-478 (2018)
A multi-dimensional approach for fractionating proteins using charged membranes
Sorci M, et al.
Biotechnology and Bioengineering, 110(6), 1704-1713 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service