Skip to Content
Merck
All Photos(1)

Documents

326534

Sigma-Aldrich

Gold

wire, diam. 0.25 mm, ≥99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
EC Number:
MDL number:
UNSPSC Code:
12141717
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9% trace metals basis

form

wire

resistivity

2.05 μΩ-cm, 0°C

diam.

0.25 mm

bp

2808 °C (lit.)

mp

1063 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Application


  • Zaleplon in Toxicological Analysis: The development of an LC-MS-MS method to detect Zaleplon along with other benzodiazepines highlights its application in forensic and clinical toxicology, providing essential data for the analysis of Z-drugs in biological samples (Sofalvi et al., 2020).

  • Study of Zaleplon′s Solvent Effects: Research on the solvent effects on Zaleplon′s absorption and fluorescence spectra aids in understanding its chemical behavior in different environments, crucial for its use in pharmaceutical formulations and biochemical studies (Divac et al., 2019).

  • Zaleplon for Insomnia Treatment via Novel Delivery Systems: The development of orodispersible film and tablets for Zaleplon delivery targets improved patient compliance and rapid onset of action in insomnia treatment, emphasizing its role in sleep disorder management (Manda et al., 2018).


Quantity

280 mg = 25 cm; 2.8 g = 250 cm

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tae-Sik Cho et al.
Journal of nanoscience and nanotechnology, 13(5), 3711-3714 (2013-07-19)
The crystallization of Au/glass ultrathin films for surface plasmon resonance (SPR) biosensor has been studied using synchrotron X-ray scattering and field emission scanning electron microscope. In films thinner than 30 nm, crystallized Au grains with [111] preferred orientation were formed
Midas touch in cardiology.
Marianna Karamanou et al.
European heart journal, 34(20), 1463-1464 (2013-07-11)
Young Joo Choi et al.
Journal of nanoscience and nanotechnology, 13(6), 4437-4445 (2013-07-19)
Gold nanorods (Au NRs) that absorb near-infrared (NIR) light have great potential in the field of nanomedicine. Photothermal therapy (PTT), a very attractive cancer therapy in nanomedicine, combines nanomaterials and light. The aim of this study was to elucidate the
Nidhi Gupta et al.
Journal of nanoscience and nanotechnology, 13(7), 4917-4924 (2013-08-02)
The preparation and photocatalytic reductive ability of a new spongy flower or cotton bud like amorphous CdS microstructures (size = 200-300 nm) having intermediate crystal phase between cubic and hexagonal structure and possesses much larger surface area ca. 119 m2
Fengxian Zheng et al.
Journal of nanoscience and nanotechnology, 13(6), 3990-3998 (2013-07-19)
Gold nanoparticles (AuNPs) are of biomedical importance, such as delivery vectors. Therefore, we used all-atom molecular dynamic simulations to study the interaction of AuNPs with cell membrane (DMPC bilayer). We observed that the AuNPs adhered spontaneously on the surface of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service