Skip to Content
Merck
All Photos(1)

Key Documents

322547

Sigma-Aldrich

1,3-Dihydro-1,3,3-trimethylspiro[2H-indole-2,3′-[3H]naphth[2,1-b][1,4]oxazine]

≥98.0% (HPLC)

Synonym(s):

Photorome I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N2O
CAS Number:
Molecular Weight:
328.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98.0% (HPLC)

mp

128-130 °C (lit.)

SMILES string

CN1c2ccccc2C(C)(C)C13Oc4ccc5ccccc5c4N=C3

InChI

1S/C22H20N2O/c1-21(2)17-10-6-7-11-18(17)24(3)22(21)14-23-20-16-9-5-4-8-15(16)12-13-19(20)25-22/h4-14H,1-3H3

InChI key

CQTRKDFIQFOAQV-UHFFFAOYSA-N

General description

1,3-Dihydro-1,3,3-trimethylspiro[2H-indole-2,3′-[3H]naphth[2,1-b][1,4]oxazine] (Photorome I) is an organic photochromic material that is used as a spirooxazine dye for use in molecular electronics. This dye changes to blue on irradiating with UV light.

Application

General applications of photochromic dye include: dosimetric materials, light controlling filters, recording films in printing processes, decorative paints, optical recording media and molecular electronic devices.1
Photorome I is a photochromic dye that can be used as a smart colorant for a variety of applications which include optical switches, printing materials, and ophthalmic lenses.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synthesis and properties of photochromic liquid-crystalline copolymers containing both spironaphthoxazine and cholesteryl groups.
Hattori H and Uryu T
Journal of Polymer Science Part A: Polymer Chemistry, 38(5), 887-894 (2000)
Interaction of a spirooxazine dye with latex and its photochromic efficiency on cellulosic paper.
Sun B, et al.
Carbohydrate Polymers, 95(1), 598-605 (2013)
NMR studies of the structure of the photoinduced forms of photochromic spironaphthoxazines.
Delbaere S, et al.
J. Chem. Soc. Perkin Trans. II, 1499-1502 (1997)
Photochromic wool fabrics from a hybrid silica coating.
Cheng T, et al.
Textile Res., 77(12), 923-928 (2007)
Cellulose nanocrystals (CNC) as carriers for a spirooxazine dye and its effect on photochromic efficiency.
Sun B, et al.
Carbohydrate Polymers, 111(8), 419-424 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service