Skip to Content
Merck
All Photos(1)

Key Documents

320293

Sigma-Aldrich

Dimethyl sulfate

≥99%

Synonym(s):

Sulfuric acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2SO2
CAS Number:
Molecular Weight:
126.13
Beilstein:
635994
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22
bp:
188 °C (lit.)
vapor pressure:
0.7 mmHg ( 25 °C)
Pricing and availability is not currently available.

vapor density

4.3 (vs air)

Quality Level

vapor pressure

0.7 mmHg ( 25 °C)

Assay

≥99%

form

liquid

autoignition temp.

923 °F

refractive index

n20/D 1.386 (lit.)

bp

188 °C (lit.)

mp

−32 °C (lit.)

density

1.333 g/mL at 25 °C (lit.)

SMILES string

COS(=O)(=O)OC

InChI

1S/C2H6O4S/c1-5-7(3,4)6-2/h1-2H3

InChI key

VAYGXNSJCAHWJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dimethyl sulfate is a powerful methylating reagent for the methylation of carboxylic acids, alcohols, phenols, lactams, oximes, hydroxylamines, and hydroperoxides.[1][2] It can be used as a reagent for the preparation of N-methyl alkyl- and aryl-substituted amines, amides, and quaternary ammonium salts. Dimethyl sulfate is also used in the methylation of sulfur-containing compounds to synthesize sulfides and sulfonium ions.[1]
It can also be used as a reagent:       
  • For the preparation of quaternary ammonium acrylic monomer as a potent antibacterial agent by the quaternization of 2-dimethylamino ethyl methacrylate.[3]       
  • Along with dimethyl sulfoxide for the synthesis of epoxides from aldehydes and ketones via formation of trimethylsulfonium cation.[4]      
  • To prepare an oxygenate additive for diesel by the methylation of glycerol.[5]

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methylation of glycerol with dimethyl sulfate to produce a new oxygenate additive for diesels
Chang J-S, et al.
Industrial & Engineering Chemistry Research, 51(2), 655-661 (2012)
Dimethyl Sulfate
Merriman G
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
Generation of trimethylsulfonium cation from dimethyl sulfoxide and dimethyl sulfate: implications for the synthesis of epoxides from aldehydes and ketones
Forrester J, et al.
Journal of the Chemical Society. Perkin Transactions 1, (18), 2289-2291 (1995)
Dimethyl Sulfate.
Merriman G.
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
Fieser and Fieser: Reagents For Organic Synthesis
Fieser, Mary and March, Jerry
Fieser and Fieser's Reagents for Organic Synthesis, 16 (1993)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service